Skip to Content
Merck
All Photos(4)

Key Documents

G5889

Sigma-Aldrich

L-Glutamic acid monosodium salt hydrate

BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%

Synonym(s):

(S)-2-Aminopentanedioic acid, L-Glutamic acid monosodium salt, Glu, MSG, Monosodium glutamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8NNaO4 · xH2O
CAS Number:
Molecular Weight:
169.11 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.75
Pricing and availability is not currently available.

product line

BioReagent

Quality Level

Assay

≥99%

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

color

white to off-white

mp

163 °C

InChI

1S/C5H9NO4.Na.H2O.H/c6-3(5(9)10)1-2-4(7)8;;;/h3H,1-2,6H2,(H,7,8)(H,9,10);;1H2;

InChI key

JYDGDQPVBSBYRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Glutamic acid/monosodium glutamate (MSG) is the sodium salt of glutamic acid.[1] It is a commonly used food additive.[1]

Application

L-Glutamic acid monosodium salt hydrate has been used:
  • to prepare glutamate stock solution in HEPES-buffered saline (HBS) for glutamate perfusions[2]
  • as a component in YNG (yeast nitrogen base glutamate) medium[3]
  • used to treat HT22 hippocampal cells to study its effects on cell viability and to examine the expression of molecules related to apoptotic cell death[4]
  • as a component of fresh media to culture neurons and to assess glutamate uptake[5]

Biochem/physiol Actions

Monosodium glutamate (MSG) is a food additive[1] and an environmental agent, that can affect inflammation.[6]
Agonist at kainate, NMDA, and quisqualate receptors; an excitatory amino acid neurotransmitter.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single-step marker switching in Schizosaccharomyces pombe using a lithium acetate transformation protocol
Brown S D and Lorenz A
Bio-protocol, 6(24) (2016)
Monosodium glutamate (MSG): a villain and promoter of liver inflammation and dysplasia
Nakanishi Y, et al.
Journal of Autoimmunity, 30(1-2), 42-50 (2008)
Microfluidic local perfusion chambers for the visualization and manipulation of synapses
Taylor A M, et al.
Neuron, 66(1), 57-68 (2010)
Food and Toxicologic Pathology: An Overview
Haschek and Rousseaux's Handbook of Toxicologic Pathology, 1051-1076 (2013)
Neuroprotective actions of noradrenaline: effects on glutathione synthesis and activation of peroxisome proliferator activated receptor delta
Madrigal J LM, et al.
Journal of Neurochemistry, 103(5), 2092-2101 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service