Skip to Content
Merck
All Photos(1)

Key Documents

G4391

Sigma-Aldrich

Gly-Arg-Gly-Asp-Ser

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H30N8O9
CAS Number:
Molecular Weight:
490.47
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97% (HPLC)

form

powder

composition

Peptide content, ≥60%

technique(s)

blocking: suitable
cell culture | mammalian: suitable

storage temp.

−20°C

SMILES string

NCC(=O)N[C@@H](CCCNC(N)=N)C(=O)NCC(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CO)C(O)=O

InChI

1S/C17H30N8O9/c18-5-11(27)23-8(2-1-3-21-17(19)20)14(31)22-6-12(28)24-9(4-13(29)30)15(32)25-10(7-26)16(33)34/h8-10,26H,1-7,18H2,(H,22,31)(H,23,27)(H,24,28)(H,25,32)(H,29,30)(H,33,34)(H4,19,20,21)/t8-,9-,10-/m0/s1

InChI key

RGNVSYKVCGAEHK-GUBZILKMSA-N

Gene Information

human ... ITGB3(3690)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Gly-Arg-Gly-Asp-Ser

General description

Gly-Arg-Gly-Asp-Ser peptide sequence is identical to the cell-binding region of fibronectin protein. Arg-Gly-Asp (RGD) region is found in various proteins, and is crucial for facilitating cell-adhesive activity.

Application

Gly-Arg-Gly-Asp-Ser has been used-
  • as an antigen to determine whether integrin α4 or α5 hinders the adhesion of day 3 postirradiation thymocytes to thymic epithelial cells, in mice
  • for the treatment of collagen IV-coated dishes to study the role of integrins α1, αv, β1, and β3 in Sca-1+ progenitor cell differentiation into SMCs (smooth muscle cells)
  • as a blocking peptide for the pre-incubation of Nisseria meningitides strains, to perform blocking experiments to study the interactions between these bacterial strains and fibronectin
  • for the pre-treatment of EL-4 cells infected by AdCMV-GFP (cytomegalovirus-green fluorescent protein), to study whether interaction with cellular integrins is essential for the transduction of adenovirus

Biochem/physiol Actions

Antagonist of integrin function
Gly-Arg-Gly-Asp-Ser inhibits metastasis in syngeneic mice or in mice lacking platelet function, when co-injected with B16-Fl0 melanoma cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PCL-PEG-based nanoparticles grafted with GRGDS peptide: preparation and surface analysis by XPS.
Vincent Pourcelle et al.
Biomacromolecules, 8(12), 3977-3983 (2007-11-28)
Yves Lecarpentier et al.
PloS one, 14(10), e0222683-e0222683 (2019-10-02)
Mesenchymal stem cells (MSCs) were obtained from human bone marrow and amplified in cultures supplemented with human platelet lysate in order to generate myofibroblasts. When MSCs were seeded in solid collagen scaffolds, they differentiated into myofibroblasts that were observed to
T Eberhard et al.
Infection and immunity, 66(4), 1791-1794 (1998-04-07)
Adhesion of Neisseria meningitidis strains to extracellular matrix (ECM) and purified matrix components was examined. Most strains bound to subendothelial ECM as well as to immobilized fibronectin and types I, III, and V collagen. Strains from healthy carriers adhered significantly
Emma L Prime et al.
Biomacromolecules, 8(8), 2416-2421 (2007-06-27)
Biodegradable polyesters such as poly(epsilon-caprolactone) (PCL) have a number of biomedical applications; however, their usage is often limited by a lack of biological functionality. In this paper, a PCL-based polymer containing pendent groups activated by 4-nitrophenyl chloroformate (NPC) and reactive
Annasara Hansson et al.
International journal of pharmaceutics, 439(1-2), 73-80 (2012-10-11)
Herein we describe the preparation of a nanoparticulate system formed from an RGD-functionalized chitosan derivative by complexation with chondroitin sulfate. These bioactive complexes were developed to promote wound healing by inducing adhesion and subsequently migration of skin cells. The particles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service