Skip to Content
Merck
All Photos(1)

Key Documents

8.02675

Sigma-Aldrich

Cyclododecanol

for synthesis

Synonym(s):

Cyclododecanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24O
CAS Number:
Molecular Weight:
184.32
MDL number:
UNSPSC Code:
12352104
EC Index Number:
217-031-2
NACRES:
NA.22

vapor pressure

<1 hPa ( 20 °C)

Quality Level

autoignition temp.

380 °C

potency

>2500 mg/kg LD50, oral (Rat)

bp

272 °C/1013 hPa

mp

75-78 °C

transition temp

flash point 138 °C

solubility

0.04 g/L

storage temp.

2-30°C

SMILES string

OC1CCCCCCCCCCC1

InChI

1S/C12H24O/c13-12-10-8-6-4-2-1-3-5-7-9-11-12/h12-13H,1-11H2

InChI key

SFVWPXMPRCIVOK-UHFFFAOYSA-N

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

280.4 °F

Flash Point(C)

138 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service