Skip to Content
Merck
All Photos(1)

Key Documents

G0503

Sigma-Aldrich

Karaya gum

Synonym(s):

Gum karaya from sterculia tree, Tragacanth Indian

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

plant fluids (Gum karaya)

form

powder

color

faint yellow to brown

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Karaya gum may be used for the development of gastro intestinal tract drug delivery vehicles, mucoadhesive sustained release drug delivery and noneffervescent floating drug delivery systems in combination with alginate.[1][2]

Other Notes

Believed to be a partially acetylated polymer of galactose, rhamnose, and glucuronic acid with a M.W. of approx. 9,500,000.
To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Singh et al.
Current drug delivery, 4(4), 269-275 (2007-11-06)
The present investigation concerns the development of floating matrix tablets of metoclopramide hydrochloride (MHCl) for improving its bioavailability by prolonging gastric residence time. Floating matrix tablets (FMT) of MHCl were prepared using the polymers guar gum (GG), karaya gum (KG)
Sadia Afrose et al.
Nutrition research (New York, N.Y.), 29(5), 350-354 (2009-06-27)
Different sources of saponins are known to have hypocholesterolemic activity with varying degrees of efficacy. We hypothesize that karaya root saponin would efficiently reduce cholesterol. The aim of this study is to examine the comparative hypocholesterolemic effect of karaya root
Baljit Singh et al.
International journal of biological macromolecules, 43(2), 142-150 (2008-05-27)
The present paper deals with the modification of the sterculia gum with methacrylic acid (MAAc) to hydrogels for use in drug delivery. The hydrogels were characterized by SEMs, FTIR and swelling studies. The release dynamics of model anti-ulcer drug (ranitidine
Kimberly A Foster et al.
International journal of pharmaceutics, 434(1-2), 406-412 (2012-06-14)
Target validation or demonstration of efficacy requires adequate in vivo exposure of tool molecules to determine their activity in order to validate the model or show the potential usefulness of the pharmacophore. Early discovery work is often carried out with
Anil Getyala et al.
Current drug delivery, 10(5), 620-629 (2013-01-05)
The aim of the work is to modify the solubility and bioavailability of Losartan potassium, by employing noneffervescent floating drug delivery (tablet dosage forms). Non-effervescent systems are a type of floating drug delivery systems, that have been used to boost

Questions

1–2 of 2 Questions  
  1. Whether Karaya Gum is obtained from tamarind seed polysaccharide ? If yes, any other seed polysaccharides also available, like amla seed, soap nut seed, basil seed etc.,

    1 answer
    1. No, this product is from the eponymously named plant, the Gum Karaya tree, not from any part of the Tamarind tree.

      Helpful?

  2. Dear Madam or Sir Is the Karaya gum suitable for food or for oral intake? Is it suitable for use in drugs or as a food supplement? Thank you!

    1 answer
    1. This product is intended for research use only and not suitable for food or oral intake. The item is not suitable for any other purpose which includes, but is not limited to, unauthorized commercial use, in vitro diagnostic use, or therapeutic use in humans or animals.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service