This product is characterized as having a low acyl content.
P8169
Phytagel™
BioReagent, suitable for plant cell culture, powder
Synonym(s):
Gellan Gum, Agar substitute gelling agent
About This Item
Recommended Products
product line
BioReagent
form
powder
technique(s)
cell culture | plant: suitable
application(s)
agriculture
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Other Notes
Reconstitution
Legal Information
also commonly purchased with this product
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.
Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.
Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.
Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.
-
Buongiorno. Vorrei sapere se il Phytagel è Gellano ad alto o a basso grado di acilazione
1 answer-
Helpful?
-
-
What is the molecular weight of one repeating unit?
1 answer-
The molecular weight is not determined. However, the constituent sugars of Phytagel are glucose, glucuronic acid and rhamnose in the molar ratio of 2:1:1.
Helpful?
-
-
What is the gelling strength of Phytagel P8169
1 answer-
The Gel Strength for Phytagel (P8169) is 700 g/cm2 (tested at 0.5%).
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service