Skip to Content
Merck
All Photos(3)

Key Documents

F6932

Sigma-Aldrich

Fucoxanthin

carotenoid antioxidant

Synonym(s):

all-trans-Fucoxanthin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H58O6
CAS Number:
Molecular Weight:
658.91
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (HPLC)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC(=O)O[C@H]1CC(C)(C)C(=C=C\C(C)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(/C)C(=O)C[C@@]23O[C@]2(C)C[C@@H](O)CC3(C)C)[C@](C)(O)C1

InChI

1S/C42H58O6/c1-29(18-14-19-31(3)22-23-37-38(6,7)26-35(47-33(5)43)27-40(37,10)46)16-12-13-17-30(2)20-15-21-32(4)36(45)28-42-39(8,9)24-34(44)25-41(42,11)48-42/h12-22,34-35,44,46H,24-28H2,1-11H3/b13-12+,18-14+,20-15+,29-16+,30-17+,31-19+,32-21+/t23-,34-,35-,40+,41+,42-/m0/s1

InChI key

SJWWTRQNNRNTPU-XJUZQKKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fucoxanthin is isolated from brown algae. It has anti-angiogenic, hepatoprotective, cardiovascular and cerebrovascular protective properties. Fucoxanthin stimulates G1 cell-cycle arrest and apoptosis in cancer cell lines.

Application

Fucoxanthin has been used to study its neuroprotective effects. It has also been used in calibration to identify the most efficient fucoxanthin producing strains of microalgae.

Biochem/physiol Actions

Xanthophyll carotenoid pigment extracted from algae. Exhibits anticancer, antioxidant, anti-obesity and anti-inflammatory properties.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bingbing Guo et al.
Pharmaceuticals (Basel, Switzerland), 14(10) (2021-10-24)
Non-alcoholic fatty liver disease (NAFLD), characterized by hepatic steatosis, is one of the most common liver diseases worldwide. So far, no definitive medical treatment has been established to treat NAFLD except for lifestyle medication. Nitzschia laevis extract (NLE), a microalgal
Ching-Shu Lai et al.
Journal of agricultural and food chemistry, 60(4), 1094-1101 (2012-01-10)
Xanthigen is a source of punicic acid and fucoxanthin derived from pomegranate seed and brown seaweed, respectively with recognized triacylglycerol-lowering effects in humans, yet the mechanism remains to be fully elucidated. The present study investigated the inhibitory effects of Xanthigen
Tim Lachnit et al.
FEMS microbiology ecology, 84(2), 411-420 (2013-01-15)
The macroalga Fucus vesiculosus carries a specific community of surface bacteria. To identify chemical compounds that possibly mediate abundance and community composition of algae-associated bacteria, we tested the effect of surface-available algal compounds on bacterial settlement and community composition under
Adah Fung et al.
Food chemistry, 136(2), 1055-1062 (2012-11-06)
This study investigated the fucoxanthin content of New Zealand (NZ) Undaria pinnatifida harvested from two locations in the Marlborough Sounds, New Zealand across its growing season. Fucoxanthin content and antioxidant properties of processed New Zealand U. pinnatifida and commercial wakame
Cheng-Ling Liu et al.
Marine drugs, 11(1), 50-66 (2013-01-10)
Cisplain, a platinum-containing anticancer drug, has been shown to enhance DNA repair and to inhibit cell apoptosis, leading to drug resistance. Thus, the combination of anticancer drugs with nutritional factors is a potential strategy for improving the efficacy of cisplatin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service