Skip to Content
Merck
All Photos(1)

Key Documents

F5379

Sigma-Aldrich

Fucosterol

≥93%, powder, antioxidant

Synonym(s):

3β-Hydroxy-5,24(28)-stigmastadiene, 5,24(28)-Stigmastadien-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H48O
CAS Number:
Molecular Weight:
412.69
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

Product Name

Fucosterol, ≥93%

Assay

≥93%

storage temp.

2-8°C

SMILES string

C\C=C(/CC[C@H](C)C1CCC2C3CC=C4CC(O)CCC4(C)C3CCC12C)C(C)C

InChI

1S/C29H48O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h7,10,19-20,23-27,30H,8-9,11-18H2,1-6H3/b21-7+

InChI key

OSELKOCHBMDKEJ-QPSGOUHRSA-N

General description

Fucosterol is a phytosterol abundantly present in algae, Ecklonia arborea and Silvetia compressa.

Application

Fucosterol has been used:
  • as an environmental contaminant to study its effect on morphological development in growing zebrafish embryos and larvae
  • to study its anxiolytic effects in mice
  • as an external standard to analyze sterols in seaweed samples

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Ikeda et al.
Journal of lipid research, 29(12), 1573-1582 (1988-12-01)
The extent and site(s) of inhibition of cholesterol absorption by plant sterols, sitosterol and fucosterol, were studied in rats. The intragastric administration of a single emulsified lipid meal containing 25 mg [3H]cholesterol and 25 mg of either sitosterol or fucosterol
J H Sheu et al.
Journal of natural products, 62(2), 224-227 (1999-03-17)
Fucosterol (1), 24xi-hydroperoxy-24-vinylcholesterol (2), 29-hydroperoxystigmasta-5,24(28)-dien-3beta-ol (3), 24-ethylcholesta-4,24(28)-dien-3-one (4), 24xi-hydroperoxy-24-ethylcholesta-4,28(29)-dien-3-one (5), 24-ethylcholesta-4,24(28)-dien-3,6-dione (6), 24xi-hydroperoxy-24-ethylcholesta-4,28(29)-dien-3,6-di one (7), 6beta-hydroxy-24-ethylcholesta-4,24(28)-dien-3-one (8), and 24xi-hydroperoxy-6beta-hydroxy-24-ethylcholesta-4,28(2 9)-dien-3-one (9) were isolated from the marine brown alga Turbinaria conoides. The structures of these compounds were established by spectral analysis.
G V Vahouny et al.
The American journal of clinical nutrition, 37(5), 805-809 (1983-05-01)
Studies have been conducted on the lymphatic absorption of sitosterol (24 alpha-ethyl cholesterol), stigmasterol (delta 22, 24 alpha-ethyl cholesterol), and fucosterol (24-ethylidine cholesterol) when administered intragastrically to rats. In addition, the effect of each sterol on absorption of endogenous cholesterol
M Shimonaka et al.
Thrombosis research, 36(3), 217-222 (1984-11-01)
In a previous study(1), it was demonstrated that one of phytosterols, sitosterol, has an ability to increase the intracellular and extracellular activities of plasminogen activator in cultured endothelial cells and that other steroids including cholesterol, 5-androsten-3 beta-ol, stigmasterol, 20(R)-propyl-5-pregnen-3 beta-ol
Cassandra D Kinch et al.
Aquatic toxicology (Amsterdam, Netherlands), 175, 286-298 (2016-04-24)
Exposure to environmental contaminants has been linked to developmental and reproductive abnormalities leading to infertility, spontaneous abortion, reduced number of offspring, and metabolic disorders. In addition, there is evidence linking environmental contaminants and endocrine disruption to abnormal developmental rate, defects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service