Skip to Content
Merck
All Photos(1)

Documents

F4799

Sigma-Aldrich

3xFLAG Peptide

≥90% (HPLC/MS), lyophilized powder

Synonym(s):

Met-Asp-Tyr-Lys-Asp-His-Asp-Gly-Asp-Tyr-Lys-Asp-His-Asp-Ile-Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys, ddddk peptide, dykddddk peptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C120H169N31O49S
Molecular Weight:
2861.87
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

product name

3X FLAG® Peptide, lyophilized powder

Assay

≥90% (HPLC/MS)

Quality Level

form

lyophilized powder

shipped in

wet ice

storage temp.

2-8°C

General description

Recommended working concentration is 100 μg/ml for elute 3X FLAG fusion proteins from the ANTI-FLAG® M2 affinity gel.
The 3X FLAG Peptide is a synthetic peptide of 23 amino acid residue. The Asp-Tyr-Lys-Xaa-Xaa-Asp motif is repeated three times in the peptide. Eight amino acids at the C-terminus make up the classic FLAG sequence (Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys).

Application

For use in competitive elution of 3X FLAG® fusion proteins from the ANTI-FLAG® M2 monoclonal antibody (F1804) in solution or bound to agarose on the ANTI-FLAG® M2 agarose affinity gel (A2220). This is achieved by Affinity Chromatography. For the gentle elution of 3X FLAG fusion proteins from the ANTI-FLAG® M2 affinity gels. 1X FLAG Peptide (F3290) will not elute 3X FLAG fusion proteins from ANTI-FLAG® affinity gels.


Learn more product details in our FLAG® application portal.

Preparation Note

To prepare a stock solution, dissolve in TBS (50 mMTris-HCl, pH 7.4, with 150 mM NaCl) at a concentration of 5 mg/ml.

Legal Information

ANTI-FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul L Colbert et al.
Oncotarget, 6(2), 953-968 (2014-12-02)
Microtubules (MTs) are components of the cytoskeleton made up of polymerized alpha and beta tubulin dimers. MT structure and function must be maintained throughout the cell cycle to ensure proper execution of mitosis and cellular homeostasis. The protein tyrosine phosphatase
Oriane Mauger et al.
Nucleic acids research, 43(3), 1869-1882 (2015-01-22)
Alternative splicing is the main source of proteome diversity. Here, we have investigated how alternative splicing affects the function of two human histone methyltransferases (HMTase): G9A and SUV39H2. We show that exon 10 in G9A and exon 3 in SUV39H2
Min Hwa Shin et al.
PLoS genetics, 12(2), e1005884-e1005884 (2016-03-02)
The inactivation of p53 creates a major challenge for inducing apoptosis in cancer cells. An attractive strategy is to identify and subsequently target the survival signals in p53 defective cancer cells. Here we uncover a RUNX2-mediated survival signal in p53
Andrew T Schiffmacher et al.
The Journal of cell biology, 215(5), 735-747 (2016-11-20)
During epithelial-to-mesenchymal transitions (EMTs), cells disassemble cadherin-based junctions to segregate from the epithelia. Chick premigratory cranial neural crest cells reduce Cadherin-6B (Cad6B) levels through several mechanisms, including proteolysis, to permit their EMT and migration. Serial processing of Cad6B by a
Tetsuya Hirata et al.
Nature communications, 9(1), 405-405 (2018-01-28)
Many eukaryotic proteins are anchored to the cell surface via the glycolipid glycosylphosphatidylinositol (GPI). Mammalian GPIs have a conserved core but exhibit diverse N-acetylgalactosamine (GalNAc) modifications, which are added via a yet unresolved process. Here we identify the Golgi-resident GPI-GalNAc

Articles

Comparison of elution techniques for small-scale protein purification of FLAG® tag proteins using anti-FLAG® M2 magnetic beads.

Protocols

Protocol for immunoprecipitation (IP) of FLAG fusion proteins using M2 monoclonal antibody 4% agarose affinity gels

Related Content

Protein purification techniques, reagents, and protocols for purifying recombinant proteins using methods including, ion-exchange, size-exclusion, and protein affinity chromatography.

Protein purification techniques, reagents, and protocols for purifying recombinant proteins using methods including, ion-exchange, size-exclusion, and protein affinity chromatography.

Protein purification techniques, reagents, and protocols for purifying recombinant proteins using methods including, ion-exchange, size-exclusion, and protein affinity chromatography.

Protein purification techniques, reagents, and protocols for purifying recombinant proteins using methods including, ion-exchange, size-exclusion, and protein affinity chromatography.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service