Skip to Content
Merck
All Photos(1)

Key Documents

F1177

Sigma-Aldrich

Fluorescein hyaluronic acid

>95%, powder

Synonym(s):

Hyaluronic Acid Fluorescein Conjugate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

Assay

>95%

form

powder

mol wt

~800,000 Da

color

orange

solubility

H2O: soluble 1 mg/mL

storage temp.

2-8°C

Application

Fluorescein hyaluronic acid has been used in the fluorescence imaging of hyaluronic acid (HA) and dextran surface coated filmsand patterned surfaces. It has also been used in confocal fluorescence microscopy to localize hyaluronic acid in collagen/HA composite gels .

Biochem/physiol Actions

Hyaluronan macromolecules (HAs) coupled to fluorescein amine is exploited to screen the hyaluronidase activity using fluorescence correlation spectroscopy (FCS) by measuring the quenching of fluorescein-labeled HA fragments. It binds on cell surface of malignant mesothelioma derived from human tumors.
Used to bind on cell surface of malignant mesothelioma derived from human tumors.

Other Notes

From cock′s comb or by fermentation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rafal Fudala et al.
Journal of photochemistry and photobiology. B, Biology, 106, 69-73 (2011-11-16)
In this report we propose a lifetime-based sensing (LBS) for the detection of hyaluronidase (HA-ase). First, we heavily label hyaluronan macromolecules (HAs) with fluorescein amine. The fluorescein labeled HA (HA-Fl) has a weak fluorescence and short fluorescence lifetime due to
Human macrophage adhesion on polysaccharide patterned surfaces
Tsai IY, et al.
Soft Matter, 7(7), 3599-3606 (2011)
Rheology and confocal reflectance microscopy as probes of mechanical properties and structure during collagen and collagen/hyaluronan self-assembly
Yang Yl and Kaufman LJ
Biophysical Journal, 96(4), 1566-1585 (2009)
Hyaluronan receptors are expressed on human malignant mesothelioma cells but not on normal mesothelial cells
Asplund T and Heldin P
Cancer research, 54(16), 4516-4523 (1994)
T Asplund et al.
Cancer research, 54(16), 4516-4523 (1994-08-15)
Hyaluronan-binding sites were demonstrated on the cell surface of three malignant mesothelioma cell lines derived from human tumors using either [3H]hyaluronan or fluorescein-tagged hyaluronan. No hyaluronan-binding activity was observed on normal human mesothelial cells. The absence of hyaluronan receptors on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service