Skip to Content
Merck
All Photos(1)

Documents

53163

Sigma-Aldrich

Hyaluronic acid sodium salt from Streptococcus equi

mol wt 750,000-1,000,000

Synonym(s):

Sodium hyaluronate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

(Streptococcus equi)

form

powder

mol wt

750,000-1,000,000

color

white

solubility

water: ~5 g/L

suitability

conforms to structure for Infrared spectrum

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C28H44N2O23.Na/c1-5(33)29-9-18(11(35)7(3-31)47-25(9)46)49-28-17(41)15(39)20(22(53-28)24(44)45)51-26-10(30-6(2)34)19(12(36)8(4-32)48-26)50-27-16(40)13(37)14(38)21(52-27)23(42)43;/h7-22,25-28,31-32,35-41,46H,3-4H2,1-2H3,(H,29,33)(H,30,34)(H,42,43)(H,44,45);/q;+1/t7-,8-,9-,10-,11-,12-,13+,14+,15-,16-,17-,18-,19-,20+,21+,22+,25-,26+,27-,28-;/m1./s1

InChI key

YWIVKILSMZOHHF-QJZPQSOGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

suitable as substrate for hyaluronidase

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dianhua Jiang et al.
Physiological reviews, 91(1), 221-264 (2011-01-21)
Accumulation and turnover of extracellular matrix components are the hallmarks of tissue injury. Fragmented hyaluronan stimulates the expression of inflammatory genes by a variety of immune cells at the injury site. Hyaluronan binds to a number of cell surface proteins
H Porsch et al.
Oncogene, 32(37), 4355-4365 (2012-10-31)
Epithelial-mesenchymal transition (EMT) is a developmental program, which can be adopted by cancer cells to increase their migration and ability to form metastases. Transforming growth factor β (TGFβ) is a well-studied inducer of EMT. We demonstrate that TGFβ potently stimulates
J Ye et al.
Eye (London, England), 26(7), 1012-1020 (2012-05-19)
To investigate the toxic effects of ethylenediaminetetraacetic acid disodium salt (EDTA), a corneal penetration enhancer in topical ophthalmic formulations, on DNA in human corneal epithelial cells (HCEs), and to investigate whether the effect induced by EDTA can be inhibited by
Melissa R Junttila et al.
Nature, 501(7467), 346-354 (2013-09-21)
Tumour formation involves the co-evolution of neoplastic cells together with extracellular matrix, tumour vasculature and immune cells. Successful outgrowth of tumours and eventual metastasis is not determined solely by genetic alterations in tumour cells, but also by the fitness advantage
Chien-Hsi Chen et al.
American journal of physiology. Heart and circulatory physiology, 306(7), H1078-H1086 (2014-02-11)
Intramyocardial injection of bone marrow mononuclear cells (MNCs) with hyaluronan (HA) hydrogel is beneficial to the ischemic heart in a rat model of myocardial infarction (MI). However, the therapeutic efficacy and safety must be addressed in large animals before moving

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service