Key Documents
EP0030048130
Eppendorf® Varitips P
pkg of 100 pieces
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
sterility:
non-sterile
packaging:
pack of 100 ea
pkg of 100 pieces
manufacturer/tradename:
Eppendorf® 0030048130
sterility
non-sterile
packaging
pack of 100 ea
pkg of 100 pieces
manufacturer/tradename
Eppendorf® 0030048130
Features and Benefits
  • Use Varitips P for aspirating 1 mL to 10 mL from beakers using the positive displacement principle
  • Use Varitips S in combination with the Eppendorf Maxitip to aspirate between 2.5 mL and 10 mL of liquid from tall, narrow-necked vessels using the air cushion principle
  • Valve for Varitips ensures drip-free dosing of liquids with high vapor pressure
  • The graduation on Varitips ensures accurate dosing of non-aqueous liquids
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)