Skip to Content
Merck
All Photos(1)

Key Documents

E3638

Sigma-Aldrich

Estazolam

Synonym(s):

8-Chloro-6-phenyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C16H11N4Cl
CAS Number:
Molecular Weight:
294.74
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

drug control

USDEA Schedule IV; Home Office Schedule 4.1; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

originator

Abbott

SMILES string

Clc1ccc-2c(c1)C(=NCc3nncn-23)c4ccccc4

InChI

1S/C16H11ClN4/c17-12-6-7-14-13(8-12)16(11-4-2-1-3-5-11)18-9-15-20-19-10-21(14)15/h1-8,10H,9H2

InChI key

CDCHDCWJMGXXRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Hypnotic; ligand for the GABAA receptor benzodiazepine modulatory site.

Features and Benefits

This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yuan Tian et al.
Journal of pharmaceutical and biomedical analysis, 47(4-5), 899-906 (2008-05-13)
A simple, rapid, sensitive and selective liquid chromatographic-tandem mass spectrometric (LC-MS/MS) method was developed and validated for the quantification of rupatadine in human plasma using estazolam as internal standard (IS). Following liquid-liquid extraction, the analytes were separated using a mobile
Masatomo Miura et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 18(5), 525-528 (2002-05-31)
A high-performance liquid chromatography (HPLC) assay was developed for the determination of estazolam in human plasma. Estazolam and alprazolam as an internal standard were detected by ultraviolet absorbance at 240 nm. Estazolam in plasma was extracted by a rapid and
H Inoue et al.
Forensic science international, 113(1-3), 367-373 (2000-09-09)
Benzodiazepines are one of the most widely prescribed drugs for the treatment of a wide spectrum of clinical disorders. They are used as anticonvulsants, anxiolytics, hypnotics or muscle relaxants with different duration of action. In this paper, a simple and
Kirkwood, C.K., et al.
Journal of the American Pharmaceutical Association, 39, 688-688 (1999)
Jiu-xi Yuan et al.
Fa yi xue za zhi, 27(2), 117-119 (2011-05-25)
To investigate the stability of estazolam in biological samples preserved in formaldehyde solution. The dog was given intragastric administration of estazolam with a dose of 37.6 mg/kg and killed 2 h later. Heart, liver, kidney and brain of the dog

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service