Skip to Content
Merck
All Photos(2)

Key Documents

E2039

Sigma-Aldrich

Chondroitinase AC from Flavobacterium heparinum

recombinant, expressed in E. coli, ≥200 units/mg protein, For Chondroitin Sulfate Analysis

Synonym(s):

Chondroitin AC lyase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

conjugate

(Glucosaminoglycan)

Assay

≥90% (SDS-PAGE)

form

lyophilized solid

specific activity

≥200 units/mg protein

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Chondroitinase AC from Flavobacterium heparinum is an enzyme that cleaves sulfated and non-sulfated polysaccharide chains with (1-4) linkages between hexosamines and glucuronic acid residues, by an elimination mechanism. The resulting oligosaccharide products are mainly disaccharides with unsaturated uronic acids. Chondroitinase AC specifically degrades chondroitin sulfates A and C, but not chondroitin sulfate B (dermatan sulfate).
Chondroitinase AC has been applied to the analysis of chondroitin sulfate in commercial samples such as dietary supplements.
Highly purified to remove interferring β-glucuronidase and protease activities for use in the hydrolysis of chondroitin sulfate pror to HPLC analysis.

Unit Definition

1 unit is defined as the amount of enzyme that will liberate 1.0 μmole per minute of unsaturated disaccharides from chondroitin sulfate A at pH 6.7 at 37 °C as measured by the change in A232. The εμΜ for the reaction product Δ-Di-4S (chondroitin sulfates A and B) is 5.1 and 5.5 for Δ-Di-6S (chondroitin sulfate C).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M Denholm et al.
European journal of pharmacology, 416(3), 213-221 (2001-04-06)
In the current study, two specific glycosaminoglycan lyases, chondroitinase AC and chondroitinase B, were utilized to examine the roles of chondroitin sulfates and dermatan sulfate in tumor metastasis and angiogenesis. Melanoma cells (SK-MEL) or endothelial cells were treated with either
Determination of Chondroitin Sulfate Content in Raw Materials and Dietary Supplements by High-Performance Liquid Chromatography with Ultraviolet Detection After Enzymatic Hydrolysis: Single-Laboratory Validation
Ji, D., et al.
Journal - Association of Official Analytical Chemists, 90(3), 659-669 (2007)
Fernanda L Paganelli et al.
International journal of antimicrobial agents, 49(3), 355-363 (2017-02-12)
Enterococcus faecium is a multidrug-resistant (MDR) nosocomial pathogen causing significant morbidity in debilitated patients. New antimicrobials are needed to treat antibiotic-resistant E. faecium infections in hospitalised patients. E. faecium incorporates lipoteichoic acid (LTA) (1,3-polyglycerol-phosphate linked to glycolipid) in its cell
Thomas N Huckerby et al.
The FEBS journal, 272(24), 6276-6286 (2005-12-13)
Chondroitin and dermatan sulfate (CS and DS) chains were isolated from bovine tracheal cartilage and pig intestinal mucosal preparations and fragmented by enzymatic methods. The oligosaccharides studied include a disaccharide and hexasaccharides from chondroitin ABC lyase digestion as well as
Studies on the enzyme chondroitinase: product structure and ion effects.
H I NAKADA et al.
Archives of biochemistry and biophysics, 94, 244-251 (1961-08-01)

Articles

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service