Skip to Content
Merck
All Photos(1)

Key Documents

C8058

Sigma-Aldrich

Chondroitinase B from Flavobacterium heparinum

lyophilized powder (with BSA as stabilizer)

Synonym(s):

Chondroitin sulfate B lyase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial (Flavobacterium heparinum)

Quality Level

conjugate

(Glucosaminoglycan)

form

lyophilized powder (with BSA as stabilizer)

shipped in

dry ice

storage temp.

−20°C

Application

Chondroitinase B from Flavobacterium heparinum has been used in a study to assess the structural characterization and antithrombin activity of dermatan sulfate. Chondroitinase B from Flavobacterium heparinum has also been used in a study to investigate the chondroitin lyase action pattern via liquid chromatography–mass spectrometry.
The enzyme from Sigma has been used for the analysis of the frequency of iduronic acid in dermatan sulfate dodecasaccharide. It has also been used to digest dermatan sufate (DS) in melanoma cells. This digestion resulted in decreased proliferation and invasiveness of tumor cells, thereby suggesting a role for DS in metastasis.

Biochem/physiol Actions

Chondroitinase B degrades only chondroitin sulphate B producing oligo- and tetra-saccharides, and an unsaturated 4-sulphated disaccharide. It has an optimum temperature of 20 °C and an optimum pH of 8.0. The enzyme activity is inhibited by 50% using 0.1 M NaCI. Co2+, Fe3+ and Ba2+ also inhibit its activity.

Unit Definition

One unit will form 0.1 μmole of unsaturated uronic acid per hr at pH 7.5 at 25°C using chondroitin sulfate B as substrate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenqing Zhang et al.
Analytical biochemistry, 385(1), 57-64 (2008-11-11)
Liquid chromatography-mass spectrometry was applied to determine the action pattern of different chondroitin lyases. Two commercial enzymes, chondroitinase ABC (Proteus vulgaris) and chondroitinase ACII (Arthrobacter aurescens), having action patterns previously determined by viscosimetry and gel electrophoresis were first examined. Next
Y M Michelacci et al.
The Biochemical journal, 151(1), 121-129 (1975-10-01)
A chondroitinase that degrades only chondroitin sulphate B was isolated from Flavobacterium heparinum, and separated from a constitutive chondroitinase AC also present in extracts of F. heparinum. The enzyme acts only on chondroitin sulphate B, producing oligo- and tetra-saccharides, plus
Yvette M Coulson-Thomas et al.
Journal of neuroscience methods, 171(1), 19-29 (2008-04-18)
Injury to the CNS of vertebrates leads to the formation of a glial scar and production of inhibitory molecules, including chondroitin sulphate proteoglycans. Various studies suggest that the sugar component of the proteoglycan is responsible for the inhibitory role of
Gerdy B ten Dam et al.
The American journal of pathology, 171(4), 1324-1333 (2007-08-25)
Chondroitin sulfate (CS) is abundantly present in the tumor stroma, and tumor-specific CS modifications might be potential targets to influence tumor development. We applied the phage display technology to select antibodies that identify these tumor-specific CS modifications. Antibody GD3G7 was
Nicola Volpi et al.
Glycobiology, 19(4), 356-367 (2008-12-06)
Glycosaminoglycans from the body of marine clam Scapharca inaequivalvis were extracted at about 0.15- 0.18 mg/g of dry tissue, composed of dermatan sulfate (DS) (approx. 74%) and heparan sulfate (26%). After treatment with nitrous acid, DS was isolated for further

Articles

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service