Skip to Content
Merck
All Photos(1)

Documents

D7815

Sigma-Aldrich

R(−)-Denopamine

≥98% (HPLC), powder

Synonym(s):

(−)-α-(3,4-Dimethoxyphenethylaminomethyl)-4-hydroxy­benzyl­alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23NO4
CAS Number:
Molecular Weight:
317.38
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: 10 mg/mL, clear
H2O: insoluble

storage temp.

2-8°C

SMILES string

COc1ccc(CCNC[C@H](O)c2ccc(O)cc2)cc1OC

InChI

1S/C18H23NO4/c1-22-17-8-3-13(11-18(17)23-2)9-10-19-12-16(21)14-4-6-15(20)7-5-14/h3-8,11,16,19-21H,9-10,12H2,1-2H3/t16-/m0/s1

InChI key

VHSBBVZJABQOSG-INIZCTEOSA-N

Gene Information

human ... ADRB1(153)

Application

R(-)-Denopamine has been used as an β-adrenergic agonist to study its effects on contractility in isolated lymphatic vessels (LVs).

Biochem/physiol Actions

Denopamine is a cardiotonic drug.
β1-adrenoceptor agonist.

Features and Benefits

This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Light sensitive.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weak arrhythmogenic property of the new cardiotonic agent denopamine in dogs: comparison with catecholamines
NARITA H, et al.
Japanese Journal of Pharmacology, 41(3), 335-344 (1986)
H Kurosawa et al.
Japanese journal of pharmacology, 72(4), 325-333 (1996-12-01)
Influence of cardiotonic agents on the prognosis of heart failure depends on the individual therapeutic agents, and favorable and unfavorable effects of these agents have been reported in clinical trials. We studied the effect of the cardiotonic agent denopamine on
Barry M Trost et al.
Organic letters, 4(16), 2621-2623 (2002-08-03)
[reaction: see text] Syntheses of variously modified ligands for the dinuclear zinc catalysts for the asymmetric aldol and nitroaldol (Henry) reactions are reported. Catalytic enantioselective nitroaldol reactions promoted by these modified ligands led to efficient syntheses of the beta-receptor agonists
Effectiveness of low dose denopamine, a beta1-adrenoceptor agonist, in a patient with vasospastic angina refractory to intensive medical treatment.
Dainari Nakajima et al.
International journal of cardiology, 108(2), 281-283 (2005-05-26)
Takeshi Ishide
Current medical research and opinion, 18(7), 407-413 (2002-12-19)
Up until now, it has been suggested that nitrate and/or calcium channel blockers were effective against variant angina pectoris. On the other hand, it is known that about 20% of variant angina pectoris was refractory to both nitrate and calcium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service