Skip to Content
Merck
All Photos(1)

Key Documents

D2532

Sigma-Aldrich

Denhardt′s Solution 50x

for Northern and Southern blotting, solution

Synonym(s):

blocking reagent, blocking solution, denhardts, hybridization buffer, hybridization solution, northern blocking solution, pre-hybridization, southern blocking solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
NACRES:
NA.25

form

solution

technique(s)

Southern blotting: suitable

foreign activity

DNAses, RNases, NICKases, endonucleases, and exonucleases, none detected

storage temp.

−20°C

General description

Denhardt′s Solution is a blocking agent routinely used to reduce non-specific binding of detection reagents to the Northern and Southern Blot membranes. This 50x concentrate can be diluted to any concentration for use in a variety of pre-hybridization and hybridization buffers.

Components

Denhardt′s Solution is 1% Bovine Serum Albumin (BSA), 1 % Ficoll, and 1 % Polyvinylpyrrolidone (PVP) in water.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Galanin receptor 3--a potential target for acute pancreatitis therapy.
Barreto SG
Neurogastroenterology and Motility, 23(3), 23(3)-23(3) (2011)
J Stokes et al.
Investigative ophthalmology & visual science, 41(7), 1629-1638 (2000-06-14)
The administration of glucocorticoids as topical or systemic medications may lead to the development of ocular hypertension through the induction of morphologic and biochemical changes in the trabecular meshwork leading to a reduction in the facility of aqueous outflow. Glucocorticoids
R E Kaye et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(18), 8542-8546 (1992-09-15)
The inhibitory effects of cyclosporin A (CsA) and FK506 on Fc epsilon receptor type I-initiated increases in cytokine mRNA and the expression of their intracellular binding proteins were studied in interleukin 3 (IL-3)-dependent, mouse bone marrow-derived mast cells (BMMCs). In
E J Mackie et al.
Journal of cell science, 103 ( Pt 3), 765-771 (1992-11-01)
The extracellular matrix glycoprotein, tenascin, is associated in vivo with mesenchyme undergoing osteogenesis and chondrogenesis, but is absent from mature bone and cartilage matrix. The expression of tenascin by osteoblastic cells in vitro has been investigated by immunoblotting and immunocytochemistry.
Gregor Obernosterer et al.
Nature protocols, 2(6), 1508-1514 (2007-06-16)
Here we describe a method for sensitive and specific histological detection of microRNAs (miRNAs) by in situ hybridization. The protocol focuses on the use of locked nucleic acids (LNAs), which are bi-cyclic RNA analogs that allow a significant increase in

Protocols

Ficoll 400® is a non-ionic polymer with high hydrophilicity and water solubility, suitable for various biological applications.

Ficoll 400® is a non-ionic polymer with high hydrophilicity and water solubility, suitable for various biological applications.

Ficoll 400® is a non-ionic polymer with high hydrophilicity and water solubility, suitable for various biological applications.

Ficoll 400® is a non-ionic polymer with high hydrophilicity and water solubility, suitable for various biological applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service