Skip to Content
Merck
All Photos(1)

Key Documents

CLS7007

Corning® Costar® Ultra-Low Attachment Multiple Well Plate

size 96 well, round bottom clear, pkg of (individually wrapped), pkg of (24/case), sterile, lid

Synonym(s):

96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, cell culture plate, tissue culture plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
polystyrene
round bottom clear
round wells
size:
96 well
surface area:
0.32 cm2 , cell growth area (cm2)
well volume:
330 μL
well working volume:
75-200 μL
wells:
96 wells

material

polystyrene
round bottom clear
round wells

sterility

sterile
sterile

feature

lid
lid
skirt
plate format: standard
well: round bottom, clear

packaging

pack of 1 (individually wrapped w/ lid)
case of 24
pkg of (24/case)
pkg of (individually wrapped)

manufacturer/tradename

Corning 7007

size

96 well

surface area

0.32 cm2 , cell growth area (cm2)

well volume

330 μL

well working volume

75-200 μL

wells

96 wells

color

clear

binding type

(Ultra-Low Attachment)

Looking for similar products? Visit Product Comparison Guide

General description

Corning Costar Ultra-Low Attachment Multiwell Plates

  • Ultra-Low attachment plates feature a covalently bound hydrogel layer that effectively inhibits cellular attachment
  • Surface minimizes protein absorption, enzyme activation, and cellular activation
  • Surface is non-cytotoxic, biologically inert, and nondegradable
  • Nonreversible lids with condensation rings to reduce contamination
  • Individual alphanumerical codes for well identification
  • Uniform footprint for ease in stacking
  • Sterilized by gamma irradiation and certified nonpyrogenic

Legal Information

Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin Izar et al.
Nature medicine, 26(8), 1271-1279 (2020-06-24)
Malignant abdominal fluid (ascites) frequently develops in women with advanced high-grade serous ovarian cancer (HGSOC) and is associated with drug resistance and a poor prognosis1. To comprehensively characterize the HGSOC ascites ecosystem, we used single-cell RNA sequencing to profile ~11,000
Nina Marie Pedersen et al.
The Journal of cell biology, 219(8) (2020-06-02)
Cancer cells break tissue barriers by use of small actin-rich membrane protrusions called invadopodia. Complete invadopodia maturation depends on protrusion outgrowth and the targeted delivery of the matrix metalloproteinase MT1-MMP via endosomal transport by mechanisms that are not known. Here

Protocols

Discover our protocol for thawing, plating, and forming hepatocyte spheroids from human spheroid-qualified plateable hepatocytes. Find the formulation for hepatocyte spheroid maintenance media.

Discover our protocol for thawing, plating, and forming hepatocyte spheroids from human spheroid-qualified plateable hepatocytes. Find the formulation for hepatocyte spheroid maintenance media.

Discover our protocol for thawing, plating, and forming hepatocyte spheroids from human spheroid-qualified plateable hepatocytes. Find the formulation for hepatocyte spheroid maintenance media.

Discover our protocol for thawing, plating, and forming hepatocyte spheroids from human spheroid-qualified plateable hepatocytes. Find the formulation for hepatocyte spheroid maintenance media.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service