Skip to Content
Merck
All Photos(1)

Key Documents

CLS3473

Corning® Costar® Ultra-Low Attachment Multiple Well Plate

size 24 well, flat bottom clear, pkg of (individually wrapped), pkg of (24/case), sterile, lid

Synonym(s):

24 well plate, cell culture plate, tissue culture plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear
flat bottom clear
polystyrene
round wells
size:
24 well
surface area:
1.9 cm2 , cell growth area (cm2)
well volume:
3.4 mL
well working volume:
0.38-0.57 mL
wells:
24 wells
Pricing and availability is not currently available.

material

clear
flat bottom clear
polystyrene
round wells

sterility

sterile
sterile

feature

lid
lid
skirt
plate format: standard
well: flat bottom, clear

packaging

pack of 1 (individually wrapped w/ lid)
case of 24
pkg of (24/case)
pkg of (individually wrapped)

manufacturer/tradename

Corning 3473

size

24 well

surface area

1.9 cm2 , cell growth area (cm2)

well volume

3.4 mL

well working volume

0.38-0.57 mL

wells

24 wells

color

clear

binding type

(ultra-low attachment)

Looking for similar products? Visit Product Comparison Guide

General description

Corning Costar Ultra-Low Attachment Multiwell Plates

  • Ultra-Low attachment plates feature a covalently bound hydrogel layer that effectively inhibits cellular attachment
  • Surface minimizes protein absorption, enzyme activation, and cellular activation
  • Surface is non-cytotoxic, biologically inert, and nondegradable
  • Nonreversible lids with condensation rings to reduce contamination
  • Individual alphanumerical codes for well identification
  • Uniform footprint for ease in stacking
  • Sterilized by gamma irradiation and certified nonpyrogenic

Legal Information

Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maxine Umeh-Garcia et al.
Cancer research, 80(3), 418-429 (2019-11-07)
miR-127 is downregulated in breast cancer, where it has been shown to suppress the proliferation, migration, and invasion of breast cancer cells. In triple-negative breast cancer (TNBC), miR-127 downregulation correlates with decreased disease-free and overall patient survival. Tumor suppressor miRNAs
Kiera L Clayton et al.
Cell host & microbe, 29(3), 435-447 (2021-02-12)
Natural killer (NK) cells are innate cytolytic effectors that target HIV-infected CD4+ T cells. In conjunction with antibodies recognizing the HIV envelope, NK cells also eliminate HIV-infected targets through antibody-dependent cellular cytotoxicity (ADCC). However, how these NK cell functions impact infected
Jin-A Lee et al.
Journal of cellular physiology, 236(5), 3946-3962 (2020-11-10)
The epigenome has an essential role in orchestrating transcriptional activation and modulating key developmental processes. Previously, we developed a library of pyrrole-imidazole polyamides (PIPs) conjugated with suberoylanilide hydroxamic acid (SAHA), a histone deacetylase (HDAC) inhibitor, for the purpose of sequence-specific

Articles

Lung organoids are valuable 3D models for human lung development and respiratory diseases. The 3dGRO™ differentiation protocol generates organoids from human iPSCs in 4 steps.

Lung organoids are valuable 3D models for human lung development and respiratory diseases. The 3dGRO™ differentiation protocol generates organoids from human iPSCs in 4 steps.

Lung organoids are valuable 3D models for human lung development and respiratory diseases. The 3dGRO™ differentiation protocol generates organoids from human iPSCs in 4 steps.

Lung organoids are valuable 3D models for human lung development and respiratory diseases. The 3dGRO™ differentiation protocol generates organoids from human iPSCs in 4 steps.

See All

Related Content

Step-by-step protocol for generating apical-out human gut organoids for microbiome, ADME/Tox, viral and gastrointestinal related disease research. See the complete organoid culture protocol.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service