Skip to Content
Merck

Skip To

CLS4580

Corning® high content imaging plate

size 96 wells, black wells (with 0.2 mm glass bottom), half area, lid, sterile

Synonym(s):

96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, imaging plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NB.13
UNSPSC Code:
41121800
Material:
black wells (with 0.2 mm glass bottom), clear bottom
Size:
96 wells
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

black wells (with 0.2 mm glass bottom), clear bottom

sterility

sterile

feature

lid, skirt, half area

packaging

pkg of 10 ea

manufacturer/tradename

Corning 4580

size

96 wells

well volume

200 μL

suitability

suitable for (cell cuture)

Application

Corning® high-content imaging plate is suitable for imaging systems to perform high-content cell-based assays. It has also been as a platform to culture the yeast cells and human mammary epithelial cell line to enable high-quality imaging during immunofluorescence (IF) microscopy.

Features and Benefits

  • Ensures a flat and optically clear surface due to high-quality glass bottom that is ideal for cell growth, decreases autofocus time, and enhances throughput
  • Anti-scratch glass
  • Well bottom flatness ensures imaging device planarity
  • Glass bottom thickness is ideal for imaging microscopy
  • Ensures the highest possible optical quality for cell-based assays due to reduced crosstalk and low background fluorescence
  • Half-area microplate consumes less reagent

Legal Information

Corning is a registered trademark of Corning, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CLS7007CLS3474CLS6511
suitability

suitable for (cell cuture)

suitability

suitable for (cell cuture)

suitability

suitable for (cell cuture)

suitability

suitable for (PCR applications)

material

black wells (with 0.2 mm glass bottom), clear bottom

material

polystyrene , round bottom clear , round wells

material

clear , flat bottom clear , polystyrene , round wells

material

clear plate, clear polycarbonate plate, conical bottom clear wells

size

96 wells

size

96 wells

size

96 wells

size

96 wells

well volume

200 μL

well volume

330 μL

well volume

360 μL

well volume

200 μL

packaging

pkg of 10 ea

packaging

pack of 1 (individually wrapped w/ lid), case of 24, pkg of (24/case), pkg of (individually wrapped)

packaging

pack of 1 (individually wrapped w/ lid), case of 24, pkg of (24/case), pkg of (individually wrapped)

packaging

pack of 1, case of 25

feature

lid, half area, skirt

feature

lid, skirt, plate format: standard, well: round bottom, clear

feature

lid, skirt, plate format: standard, well: flat bottom, clear

feature

lid: no, plate format: 96 well Model M




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library