Skip to Content
Merck
All Photos(1)

Key Documents

CLS3527

Corning® Costar® TC-Treated Multiple Well Plates

size 24 wells, flat bottom wells, case of 100 (20 Bulk Packs of 5), sterile, lid

Synonym(s):

cell culture plate, multi well plates, multiple well plates, multiwell plates, tissue culture plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
eCl@ss:
32190102
NACRES:
NB.13
material:
clear bottom
flat bottom wells
polystyrene plate
round clear wells
size:
24 wells
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
lid
skirt
Pricing and availability is not currently available.

material

clear bottom
flat bottom wells
polystyrene plate
round clear wells

description

growth area 1.9 cm2

sterility

sterile

feature

lid
skirt

packaging

case of 100 (20 Bulk Packs of 5)

manufacturer/tradename

Corning 3527

size

24 wells

well diam.

15.6 mm

well volume

3.4 mL

well working volume

0.38-0.57 mL

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Rings on lid prevent cross-contamination. All plates have a uniform footprint and a raised bead to aid stacking. Alphanumerics provide well identification. Polystyrene unless stated otherwise.

Legal Information

Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Songjie Cai et al.
Scientific reports, 10(1), 14249-14249 (2020-08-30)
Solid organ transplantation is a lifesaving therapy for patients with end-organ disease. Current immunosuppression protocols are not designed to target antigen-specific alloimmunity and are uncapable of preventing chronic allograft injury. As myeloid-derived suppressor cells (MDSCs) are potent immunoregulatory cells, we
Zili Zhang et al.
Autophagy, 16(8), 1482-1505 (2019-11-05)
Ferroptosis is a recently discovered form of programmed cell death, but its regulatory mechanisms remain poorly understood. Here, we show that the RNA-binding protein ZFP36/TTP (ZFP36 ring finger protein) plays a crucial role in regulating ferroptosis in hepatic stellate cells
Marina Koulikovska et al.
Tissue engineering. Part A, 21(5-6), 1116-1130 (2014-11-21)
Severe shortage of donor corneas for transplantation, particularly in developing countries, has prompted the advancement of bioengineered tissue alternatives. Bioengineered corneas that can withstand transplantation while maintaining transparency and compatibility with host cells, and that are additionally amenable to standardized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service