CLS3521
Corning® Netwells® accessories
carrier kit 24 mm
Synonym(s):
cell culture dishes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear plastic
sterility
non-sterile
feature
lid: no
manufacturer/tradename
Corning 3521
size
6 wells
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- These specially designed Netwell carriers and handles allow simultaneous processing of up to 6 Netwell inserts per carrier
- Each carrier kit contains eight carriers and eight handles
Lids not included
Legal Information
Corning is a registered trademark of Corning, Inc.
NETWELL is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
PloS one, 10(1), e0116766-e0116766 (2015-01-15)
Acute and chronic pain conditions are often debilitating, inflicting severe physiological, emotional and economic costs and affect a large percentage of the global population. However, the development of therapeutic analgesic agents based primarily on targeted drug development has been largely
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service