CLS3520
Corning® Netwells® accessories
carrier kit 15 mm
Synonym(s):
cell culture dishes, cell culture plates, culture dish
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear plastic
size:
12 wells
Recommended Products
material
clear plastic
sterility
non-sterile
feature
lid: no
packaging
case of 8 ea
pack of 8 ea
manufacturer/tradename
Corning 3520
size
12 wells
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- These specially designed Netwell carriers and handles allow simultaneous processing of up to 12 Netwell inserts per carrier
- Each carrier kit contains eight carriers and eight handles
Lids not included
Legal Information
Corning is a registered trademark of Corning, Inc.
NETWELL is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service