C9017
Concanavalin A from Canavalia ensiformis (Jack bean)
Sepharose™ conjugate, buffered aqueous suspension
Synonym(s):
ConA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161501
NACRES:
NA.32
Recommended Products
biological source
Canavalia ensiformis
conjugate
Sepharose™ conjugate
form
buffered aqueous suspension
contains
20% ethanol as preservative
extent of labeling
10-16 mg per mL
matrix
Sepharose 4B
matrix activation
cyanogen bromide
matrix attachment
amino
matrix spacer
1 atom
capacity
≥25 mg/mL binding capacity (thyroglobulin)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Used for the purification of glycoproteins containing mannose.
Biochem/physiol Actions
Con A is not blood group specific but has an affinity for terminal α-D-mannosyl and α-D-glucosyl residues. Ca2+ and Mn2+ ions are required for activity. Con A dissociates into dimers at pH 5.6 or below. Between pH 5.8 and pH 7.0, Con A exists as a tetramer; above pH 7.0 higher aggregates are formed. Con A exhibits mitogenic activity which is dependent on its degree of aggregation. Succinylation results in an active dimeric form which remains a dimer above pH 5.6.
Physical form
Suspension in 0.1 M acetate buffer, pH 6.0, containing 1 M NaCl, and 1 mM each of CaCl2 , MgCl2, and MnCl2
Analysis Note
Where reported, agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, containing Ca2+ and Mn2+ of a 1 mg per mL solution. This activity is the lowest concentration to agglutinate a 2% suspension of human erythrocytes after 1 hr incubation at 25 °C.
Legal Information
Sepharose is a trademark of Cytiva
related product
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 2 - Resp. Sens. 1 - Skin Sens. 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
102.2 °F
Flash Point(C)
39 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jin Lee et al.
Journal of cellular biochemistry, 112(2), 694-703 (2011-01-27)
Cadmium is one of the inflammation-related xenobiotics and has been regarded as a potent carcinogen. Gardenia jasminoides Ellis (GJE) has been used to cure inflammation in Korean folk medicine for a long time. The purpose of present study is the
Robert J Huber et al.
Matrix biology : journal of the International Society for Matrix Biology, 31(4), 271-280 (2012-03-07)
CyrA, an extracellular matrix (slime sheath), calmodulin (CaM)-binding protein in Dictyostelium discoideum, possesses four tandem EGF-like repeats in its C-terminus and is proteolytically cleaved during asexual development. A previous study reported the expression and localization of CyrA cleavage products CyrA-C45
Phil-Sun Oh et al.
Journal of cellular biochemistry, 109(1), 124-131 (2009-11-04)
This study investigated the inhibitory effect of a glycoprotein isolated from Cudrania tricuspidata Bureau (CTB glycoprotein) on di(2-ethylhexyl) phthalate (DEHP)-induced mast cell degranulation and related signaling cascade in RBL-2H3 cells. This experiment evaluated the intracellular Ca(2+) level, and the activities
Jae-Uoong Shim et al.
Inflammation, 32(4), 211-217 (2009-05-06)
Cudrania tricuspidata is one of the most omnipresent traditional herbal drugs for anti-inflammation and anti-tumor. The purpose of the present study was to determine whether the CTB glycoprotein regulates the inflammatory reaction stimulated by bisphenol A (BPA) in human mast
Sei-Jung Lee et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 87, 12-22 (2015-12-04)
This study was carried out to investigate the anti-inflammatory potentials of a 38 kDa glycoprotein isolated from Styrax japonica Siebold et al Zuccarini (SJSZ glycoprotein). We found that SJSZ glycoprotein has concentration-dependent scavenging activity against DPPH and hydroxyl radicals in the cell-free
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service