Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Concanavalin A from
Canavalia ensiformis
(Jack bean)
Synonym(s):
ConA
Products
Brand
Sigma-Aldrich (5)
Biological Source
Physical Form
Chemical Composition
Conjugate
Quality Segment
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
C2272
Concanavalin A from
Canavalia ensiformis
(Jack bean),
biotin conjugate, Type IV, lyophilized powder
Expand
View Pricing
C7642
Concanavalin A from
Canavalia ensiformis
(Jack bean),
FITC conjugate, Type IV, lyophilized powder
Expand
View Pricing
C7555
Concanavalin A from
Canavalia ensiformis
(Jack bean),
agarose conjugate, Type VI, saline suspension
Expand
View Pricing
L6397
Concanavalin A from
Canavalia ensiformis
(Jack bean),
peroxidase conjugate, lyophilized powder
Expand
View Pricing
C9017
Concanavalin A from
Canavalia ensiformis
(Jack bean),
Sepharose
™
conjugate, buffered aqueous suspension
Expand
View Pricing