Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C12H15ClO3
CAS Number:
Molecular Weight:
242.70
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
PubChem Substance ID:
EC Number:
211-277-4
Beilstein/REAXYS Number:
1913459
Form:
liquid
form
liquid
Quality Segment
color
clear colorless
density
1.14 g/mL at 25 °C (lit.)
originator
Wyeth
storage temp.
2-8°C
SMILES string
CCOC(=O)C(C)(C)Oc1ccc(Cl)cc1
InChI
1S/C12H15ClO3/c1-4-15-11(14)12(2,3)16-10-7-5-9(13)6-8-10/h5-8H,4H2,1-3H3
InChI key
KNHUKKLJHYUCFP-UHFFFAOYSA-N
Gene Information
human ... PPARA(5465)
mouse ... Ppara(19013)
Application
Clofibrate has been used:
- as a quinolinate phosphoribosyltransferase (QPRT) agonist to study its effects on hepatitis C virus (HCV) infection
- as peroxisome proliferated activated receptor (PPAR) agonist to study its effects on autophagy in rat liver cells
- as a hepatotoxicant to study its effects on gene expression in primary rat hepatocytes
Biochem/physiol Actions
Clofibrate is a peroxisome proliferated activated receptor α (PPARα) agonist. It is a fibric acid derivative and has a therapeutic effect on hypertriglyceridemia and hyperlipoproteinemia type III. Clofibrate participates in lowering the very-low-density lipoprotein (VLDL) and cholesterol levels in hyperlipoproteinemia type III patients. It facilitates the decrease of total serum bilirubin concentration in Gilbert′s syndrome.
Features and Benefits
This compound is featured on the Nuclear Receptors (PPARs) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Wyeth. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Still not finding the right product?
Explore all of our products under Clofibrate
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

