Skip to Content
Merck
All Photos(1)

Key Documents

C3784

Sigma-Aldrich

Castanospermine

≥94% (GC), BioUltra, from Castanospermum australe seeds

Synonym(s):

(1S,6S,7R,8R,8aR)-1,6,7,8-Tetrahydroxyoctahydroindolizidine, (1S,6S,7R,8R,8aR)-Octahydro-1,6,7,8-indolizinetetrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO4
CAS Number:
Molecular Weight:
189.21
Beilstein:
3588654
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

Castanospermum australe seeds

product line

BioUltra

Assay

≥94% (GC)

form

powder

mp

212-215 °C (dec.)

solubility

1 M HCl: 20 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

O[C@H]1CCN2C[C@H](O)[C@@H](O)[C@H](O)[C@@H]12

InChI

1S/C8H15NO4/c10-4-1-2-9-3-5(11)7(12)8(13)6(4)9/h4-8,10-13H,1-3H2/t4-,5-,6+,7+,8+/m0/s1

InChI key

JDVVGAQPNNXQDW-TVNFTVLESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

α-glucosidase Inhibitor

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin Pluvinage et al.
Organic & biomolecular chemistry, 7(20), 4169-4172 (2009-10-02)
The synthesis of amino-derivatives of castanospermine and australine and their characterisation as inhibitors of the exo-beta-D-glucosaminidase CsxA through enzyme kinetics and X-ray structural analysis is described.
Edward G Bowen et al.
Organic letters, 12(22), 5330-5333 (2010-10-23)
The asymmetric total synthesis of the α-glucosidase inhibitor (+)-castanospermine is reported. The central theme in our approach to this polyhydroxylated alkaloid is the simultaneous generation of the piperidine ring and the C-1/8a erythro stereodiad through the diastereoselective, oxamidation of an
M R Bartlett et al.
Immunology and cell biology, 72(5), 367-374 (1994-10-01)
The glycoprotein processing inhibitor castanospermine (CS) and the monosaccharide mannose-6-phosphate (M6P), as well as some sulfated polysaccharides (SPS), have been shown to inhibit inflammation in rat models of experimental autoimmune encephalomyelitis and adjuvant-induced arthritis. Here, the anti-inflammatory effects of these
Julien Ceccon et al.
Organic & biomolecular chemistry, 7(10), 2029-2031 (2009-05-08)
An asymmetric synthesis of (+)-castanospermine is presented in which enol ether metathesis-hydroboration/oxidation is used for stereoselective installation of the trans-trans hydroxyl groups on the piperidine ring of the alkaloid.
Matilde Aguilar-Moncayo et al.
Organic & biomolecular chemistry, 7(13), 2738-2747 (2009-06-18)
Synthesis of a panel of iso(thio)urea-type ring-modified castanospermine analogues bearing a freely mutarotating pseudoanomeric hydroxyl group results in tight-binding beta-glucosidase inhibitors with unusual binding signatures; the presence of an N-octyl substituent imparts a remarkable anomeric selectivity, promoting strong binding of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service