Skip to Content
Merck
All Photos(1)

Documents

C0399

Sigma-Aldrich

Cromolyn sodium salt

≥95%

Synonym(s):

Cromoglycate, Cromoglycic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C23H14O11Na2
CAS Number:
Molecular Weight:
512.33
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95%

form

powder

originator

Sanofi Aventis

SMILES string

[Na].OC(COc1cccc2OC(=CC(=O)c12)C(O)=O)COc3cccc4OC(=CC(=O)c34)C(O)=O

InChI

1S/C23H16O11.Na.H/c24-11(9-31-14-3-1-5-16-20(14)12(25)7-18(33-16)22(27)28)10-32-15-4-2-6-17-21(15)13(26)8-19(34-17)23(29)30;;/h1-8,11,24H,9-10H2,(H,27,28)(H,29,30);;

InChI key

UXMUBZWVLQEMRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cromolyn sodium is a mast stabilizer, which is used to treat allergies. It is a bischromone and functions as a non-steroidal anti-inflammatory drug. It inhibits the release of histamines from mast cells.

Application

Cromolyn sodium salt has been used to block mast cell activation and to inhibit histamine secretion from rodent peritoneal mast cells. It has also been used to stabilize mast cells.

Biochem/physiol Actions

Cromolyn blocks the release of histamine and other pro-inflammatory mediators from mast cells.

Features and Benefits

This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuropeptide Initiated Mast Cell Activation by Transcutaneous Electrical Acupoint Stimulation of Acupoint LI4 in Rats
Chen LZ, et al.
Scientific Reports, 8(1), 13921-13921 (2018)
Mast cells and histamine are triggering the NF-kappaB-mediated reactions of adult and aged perilymphatic mesenteric tissues to acute inflammation
Nizamutdinova IT, et al.
Aging (Albany. NY.), 8(11), 3065-3065 (2016)
Zhi Liu et al.
Journal of autoimmunity, 31(4), 331-338 (2008-10-17)
Bullous pemphigoid (BP) is a cutaneous autoimmune inflammatory disease associated with subepidermal blistering and autoantibodies against BP180, a transmembrane collagen and major component of the hemidesmosome. Numerous inflammatory cells infiltrate the upper dermis in BP. IgG autoantibodies in BP fix
Zuyi Weng et al.
PloS one, 7(3), e33805-e33805 (2012-04-04)
Mast cells are immune cells critical in the pathogenesis of allergic, but also inflammatory and autoimmune diseases through release of many pro-inflammatory cytokines such as IL-8 and TNF. Contact dermatitis and photosensitivity are skin conditions that involve non-immune triggers such
Mechanistic understanding of oral drug absorption enhancement of cromolyn sodium by an amino acid derivative
Alani AWG and Robinson JR
Pharmaceutical Research, 25(1), 48-54 (2008)

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service