Skip to Content
Merck
All Photos(2)

Documents

B5683

Sigma-Aldrich

Betaxolol hydrochloride

>98% (HPLC)

Synonym(s):

SL 75212

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H29NO3 · HCl
CAS Number:
Molecular Weight:
343.89
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

>98% (HPLC)

form

solid

storage condition

desiccated

color

white

solubility

H2O: soluble 36 mg/mL

originator

Alcon

SMILES string

Cl[H].CC(C)NCC(O)COc1ccc(CCOCC2CC2)cc1

InChI

1S/C18H29NO3.ClH/c1-14(2)19-11-17(20)13-22-18-7-5-15(6-8-18)9-10-21-12-16-3-4-16;/h5-8,14,16-17,19-20H,3-4,9-13H2,1-2H3;1H

InChI key

CHDPSNLJFOQTRK-UHFFFAOYSA-N

Gene Information

human ... ADRB1(153)

Looking for similar products? Visit Product Comparison Guide

Application

Betaxolol hydrochloride has been used as a β1 blocking agent:
  • for the preparation of crystals of (R,S)-betaxolol (BE-IV).
  • to treat mammalian cells prior cell stimulation experiments.
  • to study the solid structure of metoprolol.

Biochem/physiol Actions

Betaxolol hydrochloride has the ability to block calcium channels. Betaxolol is a selective β1 adrenoreceptor antagonist. It has neuroprotective effects on retinal neurons.

Features and Benefits

This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Alcon. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigating Differences and Similarities between Betaxolol Polymorphs
Rossi P, et al.
Crystals, 9(10), 509-509 (2019)
Yi Huang et al.
International journal of nanomedicine, 12, 1731-1745 (2017-03-11)
As a novel ion-exchange carrier with high surface area and excellent exchangeability, montmorillonite (Mt) was intercalated with betaxolol hydrochloride (BH) to form a nanocomposite and then encapsulated by liposomes (Mt-BH-LPs) for an ophthalmic drug-delivery system. The Mt-BH and Mt-BH-LPs were
João Canotilho et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 76(3-4), 395-400 (2010-04-27)
Betaxolol is a selective beta(1) receptor blocker used in the treatment of hypertension and glaucoma. A study of the betaxolol structure based on infrared spectroscopy and natural bond orbital (NBO) theory is the main aim of the present research. FTIR
Ana Franky Carvalho et al.
Neuroscience letters, 500(1), 82-85 (2011-06-23)
The cannabinoid system is known to interact with a variety of neuromodulators in the central nervous system and impacts diverse behaviors. Previous studies have demonstrated that limbic norepinephrine is a critical determinant in the behavioral expression of cannabinoid-induced aversion. The
Anders Heijl et al.
American journal of ophthalmology, 152(5), 842-848 (2011-08-17)
To analyze reduction of intraocular pressure (IOP) by argon laser trabeculoplasty (ALT) in the Early Manifest Glaucoma Trial and factors influencing the effect of such treatment. Cohort study based on 127 patients from the treatment group of the Early Manifest

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service