Skip to Content
Merck
All Photos(1)

Key Documents

B1395

Sigma-Aldrich

Anti-Bovine IgG (whole molecule) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

indirect ELISA: 1:150,000
quantitative precipitin assay: 2.0 mg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulins (Igs) belong to the immunoglobulin super-family. Each immunoglobin has two heavy (H) and two light (L) chains, held together by disulphide linkages. Heavy chain has one variable N-terminal region and three or four constant (CH1-CH4) C-terminal region. Each light chain comprises of one variable N-terminal region and a constant C-terminal region. The four classes of IgG include IgG1, IgG2, IgG3 and IgG4, among them IgG1 is most abundant.

Application

Anti-Bovine IgG (whole molecule) antibody produced in rabbit may be used for indirect ELISA at a working dilution of 1:150,000. It was used in fluorescence immunoassay for quantitative determination of IgG in bovine milk.

Biochem/physiol Actions

Deficiency of IgG1 results in hypogammaglobulinemia. IgG2 deficiency increases susceptibility to bacterial infections. IgG3 mediates effector functions. IgG4 is associated with asymptomatic infection. IgG antibody have enormous therapeutic potential and the Fc region is involved in the development of therapeutic antibody.
Digestion of IgG by papain results in generation of fragment antigen binding (Fab). Pepsin digestion of IgG results in fragment crystallisable (fc), comprises the H chain constant region. Deficiency of IgG1 results in hypogammaglobulinemia. IgG2 deficiency increases susceptibility to bacterial infections. IgG3 mediates effector functions. IgG4 is associated with asymptomatic infection. IgG antibody have enormous therapeutic potential and the Fc region is involved in the development of therapeutic antibody.
IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as preservative

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a particle concentration fluorescence immunoassay for the quantitative determination of IgG in bovine milk
Losso JN et al
Journal of Agricultural and Food Chemistry, 41, 682-686 (1993)
Structure and function of immunoglobulins
Schroeder Jr HW and Cavacini L
The Journal of Allergy and Clinical Immunology, 125, S41-S52 (2010)
Francesca Martini et al.
Nature communications, 10(1), 75-75 (2019-01-10)
The canine heartworm (Dirofilaria immitis) is a mosquito-borne parasitic nematode whose range is extending due to climate change. In a four-dimensional analysis involving HPLC, MALDI-TOF-MS and MS/MS in combination with chemical and enzymatic digestions, we here reveal an N-glycome of
Molecular properties of human IgG subclasses and their implications for designing therapeutic monoclonal antibodies against infectious diseases
Irani V, et al.
Molecular Immunology, 67 (2015)
Structure and function of immunoglobulins.
Schroeder Jr H W and Cavacini L
The Journal of Allergy and Clinical Immunology, 125(2), S41-S52 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service