Skip to Content
Merck
All Photos(1)

Key Documents

B8395

Sigma-Aldrich

Anti-Bovine IgG (whole molecule) antibody produced in rabbit

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

secondary antibodies

clone

polyclonal

contains

15 mM sodium azide

technique(s)

indirect ELISA: 1:150,000
quantitative precipitin assay: 4.0 mg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Specificity

The antiserum is determined to be immunospecific for bovine IgG.

Application

Anti-Bovine IgG (whole molecule) antibody produced in rabbit has been used as a standard to determine protein content by Bradford method using Bio-Rad protein assay dye reagent. It has also been used in several assays to develop a particle concentration fluorescence immunoassay for the quantitative determination of IgG in bovine milk.
Anti-Bovine IgG (whole molecule) antibody produced in rabbit has been used for detecting IgG antibodies in multichannel monolithic quartz crystal microbalance (MQCM).

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.
Immunoglobulins G (IgGi, IgGa) of bovine milk is capable of protecting the gut mucosa against microorganisms.

Physical form

Rabbit Anti-Bovine IgG is supplied as a liquid containing 0.1% sodium azide (see MSDS) as a preservative

Preparation Note

treated to remove lipoproteins

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of thermal protectants on the stability of bovine milk immunoglobulin G
Chao-Cheng C and Hung-Min C
Journal of Agricultural and Food Chemistry, 46(9), 3570-3576 (1998)
Development of a particle concentration fluorescence immunoassay for the quantitative determination of IgG in bovine milk
Jack NL, et al.
Journal of Agricultural and Food Chemistry, 41(4), 682-686 (1993)
Real-time multianalyte biosensors based on interference-free multichannel monolithic quartz crystal microbalance
Jaruwongrungsee K, et al.
Biosensors And Bioelectronics, 67, 576-581 (2015)
S Hashira et al.
Pediatrics international : official journal of the Japan Pediatric Society, 42(4), 337-342 (2000-09-15)
Maternal immunoglobulin G (IgG), transferred across the placenta to the fetus during intrauterine life, is an important component of the neonatal immunological defence mechanisms against infection. There is controversy with respect to differences in placental transfer of the different IgG

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service