Skip to Content
Merck
All Photos(1)

Documents

B0174

Sigma-Aldrich

AccuTaq LA 10× Buffer

10X Buffer for long and accurate PCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106306

grade

for molecular biology

Quality Level

form

liquid

packaging

vial of 0.5 mL

concentration

10 ×

technique(s)

PCR: suitable

storage temp.

−20°C

General description

AccuTaq LA 10X buffer is optimized for enhanced amplification of complex templates, when using AccuTaq long and accurate (LA) DNA polymerase.

Application

AccuTaq LA 10× Buffer has been used:

  • in PCR reaction mixture for the amplification of human BRCA1 gene.
  • as a component of the PCR mix for the amplification of the genomic DNA from Triticum aestivum
  • as a component of the PCR mix for the reverse transcription of RNA from foot and mouth disease virus (FMDV)

Legal Information

AccuTaq is a trademark of Sigma-Aldrich Co. LLC

related product

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hameer Ruparel et al.
Genome research, 14(2), 296-300 (2004-02-06)
Deletion or insertion mutations lead to a frameshift that causes misalignment between wild-type and mutated allele sequences, making it difficult to identify such mutations unambiguously by using electrophoresis-based DNA sequencing. We have previously established the feasibility of an accurate DNA
David T Hughes et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(21), 9831-9836 (2010-05-12)
The mammalian gastrointestinal (GI) tract is colonized by a complex consortium of bacterial species. Bacteria engage in chemical signaling to coordinate population-wide behavior. However, it is unclear if chemical sensing plays a role in establishing mammalian host-bacterial commensal relationships. Enterohemorrhagic
Eva-Maria Ladenburger et al.
Molecular and cellular biology, 29(13), 3605-3622 (2009-04-22)
A database search of the Paramecium genome reveals 34 genes related to Ca(2+)-release channels of the inositol-1,4,5-trisphosphate (IP(3)) or ryanodine receptor type (IP(3)R, RyR). Phylogenetic analyses show that these Ca(2+) release channels (CRCs) can be subdivided into six groups (Paramecium
Baozhong Meng et al.
The Journal of general virology, 86(Pt 5), 1555-1560 (2005-04-16)
Grapevine rupestris stem pitting-associated virus (GRSPaV), a member of the genus Foveavirus within the family Flexiviridae, is the putative causal agent of the disease Rupestris stem pitting (RSP) of grapevines. GRSPaV comprises a family of variants whose pathological characteristics have
Arunava Sarkar et al.
Biologicals : journal of the International Association of Biological Standardization, 48, 10-23 (2017-06-25)
In this study, the efficiency of binary ethyleneimine (BEI) in combination with formaldehyde (FA) and glutaraldehyde (GTA) in inactivating the Indian FMDV vaccine strains is compared. The acceptable safety of virus inactivation was faster and the inactivation rates were increased

Protocols

REDAccuTaq LA protocol offers high-fidelity amplification of long PCR fragments with direct gel loading capability.

REDAccuTaq LA protocol offers high-fidelity amplification of long PCR fragments with direct gel loading capability.

REDAccuTaq LA protocol offers high-fidelity amplification of long PCR fragments with direct gel loading capability.

REDAccuTaq LA protocol offers high-fidelity amplification of long PCR fragments with direct gel loading capability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service