Merck
Digital detection of genetic mutations using SPC-sequencing.
Genome research (2004-02-06)
Hameer Ruparel, Michael E Ulz, Sobin Kim, Jingyue Ju
ABSTRACT
Deletion or insertion mutations lead to a frameshift that causes misalignment between wild-type and mutated allele sequences, making it difficult to identify such mutations unambiguously by using electrophoresis-based DNA sequencing. We have previously established the feasibility of an accurate DNA sequencing method using solid-phase capturable (SPC) dideoxynucleotides and MALDI-TOF mass spectrometry on synthetic templates, an approach we refer to as SPC-sequencing. Here, we report the application of SPC-sequencing in characterizing frameshift mutations by using the detection of the BRCA1 gene mutations 185delAG and 5382insC as examples. In this method, Sanger DNA sequencing fragments are generated in one tube by using biotinylated dideoxynucleotides. The sequencing fragments carrying a biotin moiety at the 3' end are captured on a streptavidin-coated solid phase to eliminate excess primer, primer dimers, and false stops. Only correctly terminated DNA fragments are captured, subsequently released, and analyzed by mass spectrometry to obtain digital DNA sequencing data. This method produces distinct doublet mass peaks at each point in the mass spectrum beyond the mutation site, facilitating the accurate characterization of the mutation. We have compared SPC-sequencing with electrophoresis-based sequencing in characterizing the above BRCA1 mutations, demonstrating the significant advantage offered by SPC-sequencing for the accurate identification of frameshift mutations.
Sigma-Aldrich
AccuTaq LA 10× Buffer, 10X Buffer for long and accurate PCR