Skip to Content
Merck
All Photos(5)

Key Documents

AL0103000

Sigma-Aldrich

ALiCE® Cell Free Protein Expression Kit (Plant Cell Lysate)

An eukaryotic cell free expression system with unprecedented 3 mg/mL recombinant protein yield in batch mode and post-translational modifications enabled

Synonym(s):

Cell-Free Expression Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500
NACRES:
NA.81

form

liquid (aqueous solutions)

packaging

pkg of 6 × 200 μL (AL0103200 (ALiCE® Midi Kit))
pkg of 6 × 50 μL (AL0103050 (ALiCE® Mini Kit))
pkg of 6 × 500 μL (AL0103500 (ALiCE® Maxi Kit))

technique(s)

protein expression: suitable

shipped in

dry ice

storage temp.

−70°C

General description

Cell-free protein expression (CFPE) systems derived from crude cell extracts have been used for decades as a research tool and have introduced many attractive advantages to protein expression technology. However, a critical barrier to the adoption of CFPE systems as alternatives to cell-based approaches has been the problem of relatively low protein yields.

To address this problem, ALiCE® a novel eukaryotic cell-free expression system was created by taking advantage of plant cell biology. ALiCE® offers unprecedented yields of up to 3 mg/mL of protein in batch mode. As such, it enables you to overcome the limitations of living host systems and to obtain “difficult to produce” proteins in a matter of hours instead of weeks.

ALiCE® utilizes tobacco cell lysates, depleted of vacuoles. The system contains all factors required for in vitro transcription (RNA polymerases, NTPs) and translation reactions (ribosomes, translation initiation/elongation factors, tRNA, etc.). Using a simple protocol, both the RNA transcription and translation take place in a single tube (coupled transcription/translation).

Reactions are started by simply mixing plasmid DNA with the ALiCE® Reaction mix. The outstanding protein yields of ALiCE® are based on highly efficient protein expression over the duration of the reaction. Consequently, we recommend a reaction duration of 48 h.

ALiCE® is provided with two expression vectors, pALiCE01 and pALiCE02, which are used according to the targeting of the protein of interest. These have been specially tailored to offer excellent protein yield when used with the ALiCE® expression system. pALiCE01 is used for expression in ALiCE® cytosolic fraction. Targeting the microsomes with pALiCE02 offers the opportunity to express protein with post-translational modifications like disulfide bonds.

Application

Besides screening the ALiCE® system can also be used for larger scale expression of target proteins. Potential additional applications of the ALiCE® system include:

  • Target protein characterization
  • Protein optimization
  • Mutant screening
  • Protein-protein interactions
  • Expression analysis
  • Protein localization analysis
  • Protein structure analysis
  • Post-translational modification assays
  • Metabolomics
  • Herbicide screening

Features and Benefits

ALiCE® Cell Free Protein Expression Kit utilizes cell lysate derived from plant cells to produce recombinant protein at 3 mg/mL in batch mode.
  • High-yield: Up to 3 mg/mL expressed protein
  • Easy to use: One-step reaction protocol
  • Fast: High yield in just 48 hours
  • Flexible: Express “difficult to produce” proteins, cytotoxic proteins, etc.

Components

ALiCE® Mini Kit - AL0103050 Comprises
ALiCE® Lysate, 6 x 50 μL
Vector pALiCE01, 15 μL
Vector pALiCE02, 15 μL
ALiCE® Tube Cap, 0.6 mm hole

ALiCE® Midi Kit - AL0103200 Comprises
ALiCE® Lysate, 6 x 200 μL
Vector pALiCE01, 50 μL
Vector pALiCE02, 50 μL
ALiCE® Tube Set, 12 pcs

ALiCE® Maxi Kit - AL0103500 Comprises
ALiCE® Lysate, 6 x 500 μL
Vector pALiCE01, 50 μL
Vector pALiCE02, 50 μL
ALiCE® Tube Set, 12 pcs

Storage and Stability

  • ALiCE® Reaction (Lysate) mix: store at -70 °C
  • Vector pALiCE01: store at -20 °C to -70 °C
  • Vector pALiCE02: store at -20 °C to -70 °C
  • ALiCE® Tube caps/Tube set: store at room temperature

Legal Information

ALiCE is a registered trademark of LenioBio GmbH

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cell-free protein synthesis rapidly produces recombinant proteins without cells, utilizing translation machinery from different cell types for efficiency.

Cell-free protein synthesis rapidly produces recombinant proteins without cells, utilizing translation machinery from different cell types for efficiency.

Cell-free protein synthesis rapidly produces recombinant proteins without cells, utilizing translation machinery from different cell types for efficiency.

Cell-free protein synthesis rapidly produces recombinant proteins without cells, utilizing translation machinery from different cell types for efficiency.

Protocols

Follow these five quick steps to perform cell-free protein synthesis using the ALiCE® protein expression plant lysate kit.

Achieve high protein yields with the ALiCE® Cell-Free Protein Synthesis System that allows you to obtain “difficult to produce” proteins in a matter of hours instead of weeks.

Related Content

Read the Limited Use Label License that details how the ALiCE™ Cell Free Protein Expression System is available for in vitro research use only.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service