Skip to Content
Merck
All Photos(2)

Key Documents

A9899

Sigma-Aldrich

Antazoline hydrochloride

Synonym(s):

2-(N-Benzylanilinomethyl)-2-imidazoline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N3 · HCl
CAS Number:
Molecular Weight:
301.81
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

originator

Novartis

SMILES string

Cl.C1CN=C(CN(Cc2ccccc2)c3ccccc3)N1

InChI

1S/C17H19N3.ClH/c1-3-7-15(8-4-1)13-20(14-17-18-11-12-19-17)16-9-5-2-6-10-16;/h1-10H,11-14H2,(H,18,19);1H

InChI key

SWKDMSRRIBZZAY-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Imidazoline agonist; more potent than efaroxan in inducing insulin release from β cells; H1 histamine receptor antagonist.

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Olmos et al.
British journal of pharmacology, 127(6), 1317-1326 (1999-08-24)
This study was designed to assess the potential neuroprotective effect of several imidazol(ine) drugs and agmatine on glutamate-induced necrosis and on apoptosis induced by low extracellular K+ in cultured cerebellar granule cells. Exposure (30 min) of energy deprived cells to
M Mourtada et al.
Naunyn-Schmiedeberg's archives of pharmacology, 361(2), 146-154 (2000-02-24)
Pancreatic beta-cells express imidazoline binding sites which play a role in the regulation of insulin secretion, but it is not known whether ligands for these sites also affect other aspects of beta-cell physiology. In the present study, we have investigated
Michal M Farkowski et al.
Trials, 13, 162-162 (2012-09-13)
Rapid conversion of atrial fibrillation (AF) to sinus rhythm may be achieved by the administration of class IA, IC and III antiarrhythmic drugs or vernakalant hydrochloride. However, that treatment may be related to potential pro-arrhythmia, lack of efficacy or the
D Berdeu et al.
European journal of pharmacology, 254(1-2), 119-125 (1994-03-11)
The effects of imidazolines and derivatives were studied on insulin secretion and vascular resistance in the isolated perfused rat pancreas. On insulin secretion, two imidazoline alpha 2-adrenoceptor antagonists, efaroxan (1-100 microM) and RX821002 (10 microM), had a stimulating response; however
S C Ruckmick et al.
Journal of pharmaceutical sciences, 84(4), 502-507 (1995-04-01)
HPLC analysis of an anti-infective ophthalmic solution (Albalon-A), containing the active drugs naphazoline and antazoline, revealed a degradation peak of unknown identity. To elucidate the identity of the degradant, the active drugs were each hydrolyzed by refluxing at high pH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service