Skip to Content
Merck

Skip To

A9414

Agarose, low gelling temperature

BioReagent, Molecular Biology

Synonym(s):

3,6-Anhydro-α-L-galacto-β-D-galactan, LMP agarose, 2-Hydroxyethyl agarose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
41105317
NACRES:
NA.25
MDL number:
eCl@ss:
32110690
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

algae (red)

Quality Segment

grade

Molecular Biology

product line

BioReagent

form

powder

mp

≤65 °C

transition temp

congealing temperature 26-30 °C

gel strength

≥200 g/cm2 (1% gel)

anion traces

sulfate (SO42-): ≤0.10%

suitability

suitable for electrophoresis, suitable for molecular biology

foreign activity

DNase, RNase, NICKase, none detected

SMILES string

O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O[C@@H]4O[C@@H]5[C@H]([C@@H](OC5)[C@@H]4O)O[C@@H]6O[C@@H]([C@@H]([C@@H]([C@H]6O)O)O)CO)O)O[C@H]2[C@H]3OC[C@@H]2O[C@H]([C@H]3O)O

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

General description

Agarose is a component of agar that is naturally found in the cell wall of red algae. It is hydrophilic and has a high gelling property. It is most commonly employed for gel electrophoresis, to form porous gels to analyze DNA, RNA, or proteins by size. When a voltage is applied to the system the charged nucleic acids migrate through the agarose gel and are separated by size. The stability and gelling properties of agarose make it versitile and it can be used for many other applications in research such as, plaque and comet assays, and even as a scaffolding agent to name a few.

Application

Low-gelling agarose has been used in research:
  • for gel electrophoresis, low melting temperature is ideal for DNA extraction from gel -gel scaffold material for tissue engineering using BMSCs
  • to embed tissues in 5% agarose for immunohistology
  • for preparation of collagen-agarose co-gels to study collagen-matrix interactions in soft tissues
  • single-celled gel electrophoresis (comet assay)
  • for in-vivo 3-D imaging of zebrafish larvae immobilized on agarose strips

Features and Benefits

  • BioReagent suitable for gel electrophoresis
  • Band separation range: 200 bp - 25 kB - low gelling temperature is ideal for cell culture and viral plaque assays
  • low melting point (LMP) gel is compatible with several DNA recovery methods: phenol/chloroform extractions, recovery columns, electroelution, Beta-Agarase, and freeze/squeeze
  • Guaranteed quality tested for use in molecular biology, free of DNase and RNase

Analysis Note

The following is a list of properties associated with our agaroses:
Sulfate content - used as an indicator of purity, since sulfate is the major ionic group present.
Gel strength - the force that must be applied to a gel to cause it to fracture.
Gel point - the temperature at which an aqueous agarose solution forms a gel as it cools. Agarose solutions exhibit hysteresis in the liquid-to-gel transition - that is, their gel point is not the same as their melting temperature.
Electroendosmosis (EEO) - a movement of liquid through the gel. Anionic groups in an agarose gel are affixed to the matrix and cannot move, but dissociable counter cations can migrate toward the cathode in the matrix, giving rise to EEO. Since electrophoretic movement of biopolymers is usually toward the anode, EEO can disrupt separations because of internal convection.

Other Notes

Electroendosmosis: ≤0.10
For additional information on our range of Biochemicals, please complete this form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A0701A9045A4018
description

BioReagent, Molecular Biology

description

Type VII-A

description

BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture

description

-

form

powder

form

powder

form

powder

form

powder

gel strength

≥200 g/cm2 (1% gel)

gel strength

≥250 g/cm2 (1% gel)

gel strength

≥200 g/cm2 (1% gel)

gel strength

≥200 g/cm2 (1% gel)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

grade

Molecular Biology

grade

-

grade

-

grade

-

product line

BioReagent

product line

-

product line

BioReagent

product line

-

suitability

suitable for molecular biology, suitable for electrophoresis

suitability

-

suitability

-

suitability

-


Still not finding the right product?

Explore all of our products under Agarose, low gelling temperature


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters