Skip to Content
Merck
All Photos(7)

Key Documents

A9045

Sigma-Aldrich

Agarose, low gelling temperature

BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture

Synonym(s):

2-Hydroxyethyl agarose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41105317
NACRES:
NA.21
Pricing and availability is not currently available.

product line

BioReagent

Quality Level

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable
cell culture | plant: suitable

EEO

≤0.1

mp

≤65 °C ( at a 1.5% gel)

transition temp

congealing temperature 26-30 °C

gel strength

≥200 g/cm2 (1% gel)

anion traces

sulfate (SO42-): ≤0.10%

foreign activity

RNase and DNase free

SMILES string

O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O[C@@H]4O[C@@H]5[C@H]([C@@H](OC5)[C@@H]4O)O[C@@H]6O[C@@H]([C@@H]([C@@H]([C@H]6O)O)O)CO)O)O[C@H]2[C@H]3OC[C@@H]2O[C@H]([C@H]3O)O

InChI

1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

General description

Agarose is an algal polysaccharide. Agarose is a thermoreversible, ion-dependent gelling agent.[1] Agarose gel electrophoresis is useful for the clinical routine analyses of proteins in plasma and other body fluids.[2] It is a low gelling temperature derivative with unique gelling properties. Gels forms at <30°C, remelt at temperatures in excess of 60°C. Gels exhibit excellent clarity and are particularly useful for the preparation of media containing heat-labile materials.

Application

Agarose, a low gelling temperature derivative, is the suitable reagent for:

  • plant cell culture studies
  • insect cell culture studies
  • cell culture studies

It may be used for the following studies:
  • Recovery of defined RNA and DNA fractions after electrophoretic separation.[3]
  • Cytochemical staining procedure to investigate the succinate dehydrogenase (SDH) activity in pre-ovulatory mouse oocytes.[4]
  • Purification of RNA in Caenorhabditis elegans by electrophoresis.[5]

Analysis Note

The following is a list of properties associated with our agaroses:
Sulfate content - used as an indicator of purity, since sulfate is the major ionic group present.
Gel strength - the force that must be applied to a gel to cause it to fracture.
Gel point - the temperature at which an aqueous agarose solution forms a gel as it cools. Agarose solutions exhibit hysteresis in the liquid-to-gel transition - that is, their gel point is not the same as their melting temperature.
Electroendosmosis (EEO) - a movement of liquid through the gel. Anionic groups in an agarose gel are affixed to the matrix and cannot move, but dissociable counter cations can migrate toward the cathode in the matrix, giving rise to EEO. Since electrophoretic movement of biopolymers is usually toward the anode, EEO can disrupt separations because of internal convection.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katherine A Pillman et al.
The EMBO journal, 37(13) (2018-06-07)
Members of the miR-200 family are critical gatekeepers of the epithelial state, restraining expression of pro-mesenchymal genes that drive epithelial-mesenchymal transition (EMT) and contribute to metastatic cancer progression. Here, we show that miR-200c and another epithelial-enriched miRNA, miR-375, exert widespread
A Fire et al.
Nature, 391(6669), 806-811 (1998-03-05)
Experimental introduction of RNA into cells can be used in certain biological systems to interfere with the function of an endogenous gene. Such effects have been proposed to result from a simple antisense mechanism that depends on hybridization between the
Patrick M Ferree et al.
Scientific reports, 9(1), 12194-12194 (2019-08-23)
Males of hymenopteran insects, which include ants, bees and wasps, develop as haploids from unfertilized eggs. In order to accommodate their lack of homologous chromosome pairs, some hymenopterans such as the honeybee have been shown to produce haploid sperm through
Agarose gel electrophoresis.
Johansson B G.
Scandinavian Journal of Clinical and Laboratory Investigation, 29(S124), 7-19 (1972)
Nariko Arimura et al.
Science advances, 6(36) (2020-09-13)
For normal neurogenesis and circuit formation, delamination of differentiating neurons from the proliferative zone must be precisely controlled; however, the regulatory mechanisms underlying cell attachment are poorly understood. Here, we show that Down syndrome cell adhesion molecule (DSCAM) controls neuronal

Questions

1–6 of 6 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Can Agarose, low gelling temperature, be autoclaved?

    1 answer
    1. This agarose can be autoclaved.

      Helpful?

  4. What is the gel strength of Agarose, low gelling temperature?

    1 answer
    1. We have a specification of not less than 200 g/cm2 for a 1.0% agarose gel.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. How do I use Agarose, low gelling temperature?

    1 answer
    1. This product forms a suspension in nearly any aqueous buffer. Heating will melt the agar, which, upon cooling, will form a gel.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service