Skip to Content
Merck
All Photos(1)

Key Documents

A9171

Sigma-Aldrich

Anti-Chicken IgY (IgG) (whole molecule)−Alkaline Phosphatase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Rabbit Anti-Chicken IgY (IgG) (whole molecule)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:35,000
dot blot: 1:100,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:40
western blot (chemiluminescent): suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin Y (IgY) is also known as egg yolk immunoglobulin and is found mainly in birds, amphibians and reptiles. IgY cannot activate mammalian complement system and cannot bind to mammalian rheumatoid factors, and protein A or G. IgY facilitates an essential role in veterinary science, functional food and bio-products. It also has a crucial role in controlling infectious diseases of bacterial and viral origin.

Specificity

Anti-Chicken IgY (IgG) (whole molecule)-Alkaline Phosphatase antibody is specific for chicken IgY (IgG).

Immunogen

IgY (IgG) isolated from pooled normal chicken serum
IgY (IgG) isolated from pooled normal chicken serum.

Application

Anti-Chicken IgY (IgG) (whole molecule) Alkaline Phosphatase antibody produced in rabbit has been used in:
  • immunoblotting
  • indirect enzyme linked immunosorbent assay (ELISA)
  • immunoglobulin assay
  • dot Blot
  • immunohistology

Biochem/physiol Actions

Chicken IgY is the major antibody found in chicken serum. Chickens generate a comparatively enhanced antibody response to mammalian antigenic proteins. These immunoglobulins do not bind to Fc receptors present in mammals and bacteria, and also do not interact with rheumatoid factors. Thus anti-chicken antibodies are useful analytical tools for various research applications.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2 and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naresh Loudya et al.
Genome biology, 22(1), 151-151 (2021-05-13)
The developmental gradient in monocot leaves has been exploited to uncover leaf developmental gene expression programs and chloroplast biogenesis processes. However, the relationship between the two is barely understood, which limits the value of transcriptome data to understand the process
Variable exposure and immunological response to Lyme disease Borrelia among North Atlantic seabird species
Staszewski V, et al.
Proceedings of the Royal Society of London. Series B, Containing Papers of a Biological Character. Royal Society (Great Britain), 275(1647), 2101-2109 (2008)
Effect of dietary protein concentrates on the incidence of subclinical necrotic enteritis and growth performance of broiler chickens
Palliyeguru MWCD
Poultry Science, 89(1), 34-43 (2010)
Ana Barbosa Marcondes de Mattos et al.
Frontiers in physiology, 14, 1252470-1252470 (2024-01-04)
Abdominal aortic aneurysm (AAA) is a life-threatening vascular disease for which surgical or endovascular repair are the only currently available therapeutic strategies. The development of AAA involves the breakdown of elastic fibers (elastolysis), infiltration of inflammatory cells, and apoptosis of
Minh-Uyen Thi Le et al.
Analytical chemistry, 94(26), 9297-9305 (2022-06-14)
The importance of multi-omic-based approaches to better understand diverse pathological mechanisms including neurodegenerative diseases has emerged. Spatial information can be of great help in understanding how biomolecules interact pathologically and in elucidating target biomarkers for developing therapeutics. While various analytical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service