Skip to Content
Merck
All Photos(6)

Documents

SAB2702218

Sigma-Aldrich

Monoclonal Anti-6X His tag antibody produced in mouse

clone GT359, affinity isolated antibody

Synonym(s):

Mouse Anti-Hexa Histidine tag

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

GT359, monoclonal

form

buffered aqueous solution

species reactivity

human, mouse

concentration

1mg/mL

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: 500-3000

isotype

IgG3

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

6X His is a histidine tag that consists of a series of histidine residues at the amino or carboxy terminus of a protein.

Immunogen

The immunogen used to generate this antibody corresponds to 6X His tag

Application

Suggested starting dilutions are as follows: ELISA: 1:1000-1:10000, ICC/IF: 1:100-1:2000, IP: 1:100-1:500, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Monoclonal Anti-6X His tag antibody produced in mouse has been used in Co-Immunoprecipitation (Co-IP) and immunoblotting.

Biochem/physiol Actions

6X His tag does not affect the structure and function of protein and can be used in antibody production. It is also used to purify proteins under native or denaturing form.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Other Notes

Purification: Affinity purified by Protein G

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohaddeseh Nasir Shirazi et al.
Iranian journal of biotechnology, 22(1), e3602-e3602 (2024-06-03)
Uricase or urate oxidase, as a therapeutic enzyme, is extensively applied to oxidize accumulated uric acid in the body to soluble form to treat related illnesses. This study was conducted with the aim of searching for potential sources of uricase-producing
SKP2-and OTUD1-regulated non-proteolytic ubiquitination of YAP promotes YAP nuclear localization and activity.
Yao F, et al.
Nature Communications, 9(1), 2269-2269 (2018)
Jordan Demone et al.
PloS one, 17(12), e0277668-e0277668 (2022-12-15)
Agroinfiltration is a method used in biopharming to support plant-based biosynthesis of therapeutic proteins such as antibodies and viral antigens involved in vaccines. Major advantages of generating proteins in plants is the low cost, massive scalability and the rapid yield
Erratum: Engineering of the Recombinant Expression and PEGylation Efficiency of the Therapeutic Enzyme Human Thymidine Phosphorylase.
Frontiers Production Office
Frontiers in Bioengineering and Biotechnology, 10, 878674-878674 (2022)
James S Terry et al.
Virology, 558, 28-37 (2021-03-15)
To help fight COVID-19, new molecular tools specifically targeting critical components of the causative agent of COVID-19, SARS-Coronavirus-2 (SARS-CoV-2), are desperately needed. The SARS-CoV-2 nucleocapsid protein is critical for viral replication, integral to viral particle assembly, and a major diagnostic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service