Skip to Content
Merck
All Photos(1)

Key Documents

A8351

Sigma-Aldrich

Ampicillin sodium salt

BioXtra, suitable for cell culture

Synonym(s):

D-(−)-α-Aminobenzylpenicillin sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N3NaO4S
CAS Number:
Molecular Weight:
371.39
Beilstein:
4119211
EC Number:
MDL number:
UNSPSC Code:
51281716
PubChem Substance ID:
NACRES:
NA.76
Pricing and availability is not currently available.

product line

BioXtra

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white to off-white

mp

215 °C (dec.) (lit.)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].CC1(C)SC2[C@H](NC(=O)[C@H](N)c3ccccc3)C(=O)N2[C@H]1C([O-])=O

InChI

1S/C16H19N3O4S.Na/c1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8;/h3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23);/q;+1/p-1/t9-,10-,11+,14-;/m1./s1

InChI key

KLOHDWPABZXLGI-YWUHCJSESA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Application

Ampicillin is a semi-synthetic derivative of penicillin that functions as a broad-spectrum antibiotic. It has been used to study antibiotic resistance and penetration limitations, the synergy between multiple antibiotics, certain bloodstream infections, and has been used to develop PCR assays to detect resistance genes in cerebrospinal fluid.

Biochem/physiol Actions

Mode of Action: This is a ß-lactam antibiotic that inhibits bacterial cell-wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane.

Mode of Resistance: Administration with ß-lactamase cleaves the ß-lactam ring of Ampicillin and inactivates it.

Antimicrobial Spectrum: Includes both gram-positive (similar to benzylpenicillin) and gram-negative bacteria (similar to tetracyclines and chloramphenicol.

Caution

This product has been reported stable as supplied at 25°C at 43% and 81% relative humidity for six weeks. Additional studies have shown that the stability of Ampicillin in solution is a function of pH, temperature and the identity of the buffer. It′s activity is quickly lost when stored above pH 7. Optimal storage conditions are suggested as 2-8°C, and pH 3.8-5 where its activity was retained at 90%+ for a week.

Preparation Note

Ampicillin is reported as slightly soluble in water, practically insoluble in alcohol, chloroform, ether and fixed oils but soluble in dilute acids or bases. The solution should not be autoclaved; a stock solution should be sterilized through filtration and stored frozen, where it will be stable for months.

Other Notes

Keep container tightly closed in a dry and well-ventilated place, hygroscopic.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1A - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jaroslav Slamecka et al.
Cell cycle (Georgetown, Tex.), 17(3), 330-347 (2017-11-17)
Fetal stem cells are a unique type of adult stem cells that have been suggested to be broadly multipotent with some features of pluripotency. Their clinical potential has been documented but their upgrade to full pluripotency could open up a
Jianhua Yin et al.
PloS one, 8(3), e60460-e60460 (2013-04-05)
Shewanella oneidensis is a facultative anaerobic γ-proteobacterium possessing remarkably diverse respiratory capacities for reducing various organic and inorganic substrates. As a veteran research model for investigating redox transformations of environmental contaminants the bacterium is well known to be a naturally
Nuria Fernández-Hidalgo et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 56(9), 1261-1268 (2013-02-09)
The aim of this study was to compare the effectiveness of the ampicillin plus ceftriaxone (AC) and ampicillin plus gentamicin (AG) combinations for treating Enterococcus faecalis infective endocarditis (EFIE). An observational, nonrandomized, comparative multicenter cohort study was conducted at 17
Sabine Renggli et al.
Journal of bacteriology, 195(18), 4067-4073 (2013-07-10)
Bactericidal antibiotics kill by different mechanisms as a result of a specific interaction with their cellular targets. Over the past few years, alternative explanations for cidality have been proposed based on a postulated common pathway, depending on the intracellular production
Tonia Zangari et al.
Infection and immunity, 81(5), 1562-1574 (2013-02-27)
In May 2011, a large food-borne outbreak was traced to an unusual O104:H4 enteroaggregative Escherichia coli (EAEC) strain that produced Shiga toxin (Stx) type 2 (Stx2). We developed a mouse model to study the pathogenesis and treatment for this strain

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service