Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ampicillin sodium salt
Synonym(s):
D-(−)-α-Aminobenzylpenicillin sodium salt
Empirical Formula (Hill Notation):
C
16
H
18
N
3
NaO
4
S
CAS Number:
69-52-3
Molecular Weight:
371.39
EC Number:
200-708-1
Products
Product Category
antibiotics (3)
Brand
Sigma-Aldrich (3)
Biological Source
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Activity Spectrum
Quality Segment
Application
Mode of Action
Optical Isomer
Special Grade
Biotechnology Performance Certified (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A0166
Ampicillin sodium salt,
powder or crystals, BioReagent, suitable for cell culture
Expand
View Pricing
A9518
Ampicillin sodium salt,
Expand
View Pricing
A8351
Ampicillin sodium salt,
BioXtra, suitable for cell culture
Expand
View Pricing