Skip to Content
Merck
All Photos(4)

Key Documents

A6272

Sigma-Aldrich

Albumin from rat serum

lyophilized powder, ≥96% (agarose gel electrophoresis)

Synonym(s):

RSA, Rat albumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27
Pricing and availability is not currently available.

biological source

rat

Quality Level

Assay

≥96% (agarose gel electrophoresis)

form

lyophilized powder

technique(s)

ELISA: suitable
tissue culture: suitable
western blot: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

rat ... ALB(24186)

Looking for similar products? Visit Product Comparison Guide

General description

Albumin from rat serum (RSA) comprises an N-terminal signal and propeptide sequence.[1] RSA sequence has a single tryptophan residue.[2]

Application

Albumin from rat serum has been used:
  • in sulfo-4-(N-maleimidomethyl)cyclohexane-1-carboxylic,acid 3-sulfo-N-hydroxy-succinimide ester sodium salt (SMCC) derivatization studies[3]
  • as a component of dispersion media for the suspension of multi-walled carbon nanotubes (MWCNTs)[4]
  • as a macromolecular probe in microvascular solute flux measurements[5]

Biochem/physiol Actions

Albumin from rat serum (RSA) mediates the transport of exogenous ligands. It is useful in fluorescence spectroscopy measurements with small-molecule ligands.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A K Vidanapathirana et al.
Journal of nanomedicine & nanotechnology, 5(3) (2014-06-01)
Multi-walled carbon nanotubes (MWCNTs) are increasingly used in industry and in nanomedicine raising safety concerns, especially during unique life-stages such as pregnancy. We hypothesized that MWCNT exposure during pregnancy will increase vascular tissue contractile responses by increasing Rho kinase signaling.
Gabriel Zazeri et al.
International journal of molecular sciences, 20(12) (2019-06-20)
The bioactive piperine (1-piperoyl piperidine) compound found in some pepper species (Piper nigrum linn and Piper sarmentosum Roxb) has been shown to have therapeutic properties and to be useful for well-being. The tests used to validate these properties were performed
Theo Hagg
Methods in molecular biology (Clifton, N.J.), 399, 167-180 (2008-03-04)
Neurotrophic factors are among the most potent neuroprotective and neuroregenerative agents known. However, they cross the adult mammalian blood-brain barrier very poorly and can have serious peripheral side effects. These problems can be solved by using chronic infusions with small
Sejal S Hall et al.
Biotechnology progress, 23(3), 749-754 (2007-05-02)
Peptide ligands capable of mediating nanoparticle adhesion to human red blood cells (RBCs) were identified from a large bacterial display peptide library. Peptides were displayed on the surface of fluorescent Escherichia coli, enabling quantitative measurement of RBC binding and high-throughput
Joshua P Scallan et al.
The Journal of physiology, 591(20), 5071-5081 (2013-07-31)
Atrial and brain natriuretic peptides (ANP and BNP, respectively) are cardiac hormones released into the bloodstream in response to hypervolaemia or fluid shifts to the central circulation. The actions of both peptides include natriuresis and diuresis, a decrease in systemic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service