A6412
Aflatoxin M1–BSA Conjugate
Synonym(s):
AFM1-BSA conjugate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
powder
Quality Level
extent of labeling
4-8 mol aflatoxin M1 per mol BSA (Lowry)
storage temp.
2-8°C
General description
Aflatoxin M1 is a hydroxylated metabolite ofaflatoxin B1. It is found in milk secreted by mammary glands of lactatinganimals, humans, and milk products.
Application
Aflatoxin M1–BSA Conjugate has been used in the indirect competitive enzyme-linked immunosorbent assay (ic-ELISA) with non-labeled aflatoxin M1 and anti-AFM1 antibody.
Biochem/physiol Actions
Aflatoxin M1 is a mycotoxin that elicits both hepatotoxic and hepatocarcinogenic effects leading to immunosuppression in animals. The bioconjugate of aflatoxin M1 (AFM1) with bovine serum albumin (BSA) is useful in the generation of AFM1 specific antibodies which may have immunoassay applications.
Warning
Possibly carcinogenic.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Carc. 1B
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of agricultural and food chemistry, 50(4), 933-937 (2002-02-07)
High-titer rabbit polyclonal antibodies to aflatoxin M(1) (AFM1) were produced by utilizing AFM1-bovine serum albumin (BSA) conjugate as an immunogen. An indirect competitive enzyme-linked immunosorbent assay was standardized for estimating AFM1 in milk and milk products. To avoid the influence
Jundishapur journal of microbiology, 8(4), e16850-e16850 (2015-06-03)
Aflatoxins are the most extensively studied group of mycotoxins produced by molds, especially the Aspergillus group, which are highly toxic to animals and humans. Since immunoassay is a simple and rapid method for the analysis of many toxic substances in
Talanta, 77(1), 138-143 (2008-09-23)
Five different clones of antibodies developed against the aflatoxin M(1) were investigated by using the classical indirect and direct competitive Enzyme-Linked Immunosorbent Assay (ELISA) formats, and also the direct competitive ELISA based on the use of the superparamagnetic nanoparticles. The
Aflatoxin M1 in milk and dairy products, occurrence and recent challenges: A review
Trends in Food Science & Technology, 46 (1) (2015)
Frontiers in microbiology, 7, 2170-2170 (2017-02-02)
The aflatoxin producing fungi, Aspergillus spp., are widely spread in nature and have severely contaminated food supplies of humans and animals, resulting in health hazards and even death. Therefore, there is great demand for aflatoxins research to develop suitable methods
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service