Skip to Content
Merck

Skip To

A5575

Methyl δ-aminolevulinate hydrochloride

≥98% (TLC)

Synonym(s):

5-Amino-4-oxopentanoic acid methyl ester, 5-Aminolevulinic acid methyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H11NO3 · HCl
CAS Number:
Molecular Weight:
181.62
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352209
EC Number:
279-151-1
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Methyl δ-aminolevulinate hydrochloride, ≥98% (TLC)

Quality Level

assay

≥98% (TLC)

form

powder

color

white

storage temp.

2-8°C

SMILES string

Cl.COC(=O)CCC(=O)CN

InChI

1S/C6H11NO3.ClH/c1-10-6(9)3-2-5(8)4-7;/h2-4,7H2,1H3;1H

InChI key

UJYSYPVQHFNBML-UHFFFAOYSA-N

Biochem/physiol Actions

Methyl δ-aminolevulinate (methyl-ALA, MAL) and 5-Aminolevulinic acid (5-ALA) are commonly used in photosentizer reagents for photodynamic therapy to treat conditions such as Acne vulgaris and hypertrophic scarring.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A3785A7793672629
form

powder

form

powder

form

powder

form

solid

assay

≥98% (TLC)

assay

≥98%

assay

≥98%

assay

≥96.5% (HPLC), 97%

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

color

white

color

-

color

-

color

-

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

100


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Photodynamic therapy for acne vulgaris: a critical review from basics to clinical practice: part I. Acne vulgaris: when and why consider photodynamic therapy?
Sakamoto FH, Lopes JD, Anderson RR.
Journal of the American Academy of Dermatology, 63, 193-194 (2010)
Nicola Bruscino et al.
Photodermatology, photoimmunology & photomedicine, 27(6), 334-335 (2011-11-19)
The case we report is about a female patient, 69 years old, who had a hypertrophic scarring on the right cheek because of a bite by her dog. She had attempted many types of topical and intralesional treatments but without
Yoojin Lee et al.
Seminars in cutaneous medicine and surgery, 30(4), 199-209 (2011-11-30)
Photodynamic therapy (PDT) involves the activation of a photosensitizing drug, which preferentially localizes to diseased skin, by irradiation with light to cause selective cytotoxic damage. Since its discovery in the early 20th century and the development of topical photosensitizers 2



Global Trade Item Number

SKUGTIN
A5575-100MG04061832421131
A5575-1G04061826055540

Questions

Reviews

No rating value

Active Filters