Skip to Content
Merck
All Photos(1)

Key Documents

A4330

Sigma-Aldrich

4′-Aminomethyltrioxsalen hydrochloride

Synonym(s):

4′-Aminomethyl-4,5′,8-trimethylpsoralen hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15NO3 · HCl
CAS Number:
Molecular Weight:
293.75
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

solubility

H2O: 1 mg/mL
DMSO: 2 mg/mL

storage temp.

2-8°C

SMILES string

CC1=CC(=O)Oc2c(C)c3oc(C)c(CN)c3cc12

InChI

1S/C15H15NO3/c1-7-4-13(17)19-14-8(2)15-11(5-10(7)14)12(6-16)9(3)18-15/h4-5H,6,16H2,1-3H3

InChI key

WBIICVGYYRRURR-UHFFFAOYSA-N

Application

4′-Aminomethyltrioxsalen hydrochloride inactivates DNA and RNA viruses, including HIV-1. 4′-Aminomethyltrioxsalen hydrochloride can covalently bind to nucleic acids when irradiated with UV light.

Biochem/physiol Actions

4′-Aminomethyltrioxsalen hydrochloride is used to inactivate DNA and RNA viruses, including HIV-1, by nucleic acid cross-linking followed by UV irradiation.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Grass et al.
Blood, 91(6), 2180-2188 (1998-04-16)
A photochemical treatment (PCT) process using a novel psoralen and long wavelength ultraviolet light (UVA, 320-400 nm) has been developed to inactivate bacteria and viruses in platelet concentrates. This study evaluated the efficacy of PCT for inactivation of leukocytes that
H Du et al.
RNA (New York, N.Y.), 7(1), 133-142 (2001-02-24)
Base pairing between the 5' end of U1 snRNA and the conserved 5' splice site of pre-mRNA is important for commitment complex formation in vitro. However, the biochemical mechanisms by which pre-mRNA is initially recognized by the splicing machinery is
W S Groene et al.
Journal of virological methods, 38(1), 93-102 (1992-07-01)
The use of the synthetic psoralen 4'-aminomethyl-4,5',8-trimethylpsoralen hydrochloride (AMT) is described for the inactivation of infectious rotavirus, a member of the viral family Reoviradae with a double-stranded RNA genome. This method not only provides complete inactivation of the virus but
M Beltrame et al.
The EMBO journal, 11(4), 1531-1542 (1992-04-01)
It has long been known that U3 can be isolated hydrogen bonded to pre-ribosomal RNAs, but the sites of interaction are poorly characterized. Here we show that yeast U3 can be cross-linked to 35S pre-rRNA both in deproteinized extracts and
A A Oroskar et al.
Free radical biology & medicine, 20(5), 751-756 (1996-01-01)
The ability of scavengers of hydroxyl radical (OH radical) to modulate the photosensitized relaxation (induction of the first single-strand break) of supercoiled plasmid DNA with UVA photoactivated 4'-aminomethyl-4,5',8-trimethylpsoralen was examined by comparing the dose reduction factor (DRF: the ratio of

Articles

DNA damage and repair mechanism is vital for maintaining DNA integrity. Damage to cellular DNA is involved in mutagenesis, the development of cancer among others.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service