Skip to Content
Merck
All Photos(1)

Key Documents

A2407

Sigma-Aldrich

Adrenocorticotropic Hormone Fragment 1-17 human, rat

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C95H145N29O23S
CAS Number:
Molecular Weight:
2093.41
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32
biological source:
synthetic (organic)
form:
powder
Assay:
≥97% (HPLC)

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥97% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCC[C@H](NC(=O)[C@H](CO)NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@@H](N)CO)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](Cc2c[nH]cn2)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N6CCC[C@H]6C(=O)N[C@@H](C(C)C)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C95H145N29O23S/c1-53(2)78(91(144)109-49-75(128)111-62(22-9-12-35-96)81(134)113-63(23-10-13-36-97)82(135)117-68(93(146)147)26-16-39-106-95(102)103)123-90(143)74-27-17-40-124(74)92(145)67(24-11-14-37-98)112-76(129)48-108-80(133)71(44-56-46-107-61-21-8-7-20-59(56)61)120-83(136)64(25-15-38-105-94(100)101)114-86(139)70(42-54-18-5-4-6-19-54)119-88(141)72(45-57-47-104-52-110-57)121-84(137)65(32-33-77(130)131)115-85(138)66(34-41-148-3)116-89(142)73(51-126)122-87(140)69(118-79(132)60(99)50-125)43-55-28-30-58(127)31-29-55/h4-8,18-21,28-31,46-47,52-53,60,62-74,78,107,125-127H,9-17,22-27,32-45,48-51,96-99H2,1-3H3,(H,104,110)(H,108,133)(H,109,144)(H,111,128)(H,112,129)(H,113,134)(H,114,139)(H,115,138)(H,116,142)(H,117,135)(H,118,132)(H,119,141)(H,120,136)(H,121,137)(H,122,140)(H,123,143)(H,130,131)(H,146,147)(H4,100,101,105)(H4,102,103,106)/t60-,62-,63-,64-,65-,66-,67-,68-,69-,70-,71-,72-,73-,74-,78-/m0/s1

InChI key

MXNMAFWEFLFAJN-QBQLSIIBSA-N

Gene Information

human ... POMC(5443)

Amino Acid Sequence

Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg

Biochem/physiol Actions

ACTH fragment with moderate adrenocorticotropic and melanocyte-stimulating activities.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Skin POMC peptides. Their binding affinities and activation of the human MC1 receptor.
M Tsatmalia et al.
Annals of the New York Academy of Sciences, 885, 466-469 (2000-05-19)
M Terzolo et al.
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology, 5(2), 83-94 (1991-06-01)
The aim of the present study was to analyze the effects of exogenous melatonin (MT) upon pituitary and adrenal responsiveness to releasing hormones in different phases of the menstrual cycle. We evaluated the response of FSH and LH to 100
P Paccotti et al.
Chronobiologia, 15(4), 279-287 (1988-10-01)
We evaluated the effect of an acute oral administration of 2 dosages (100 and 1 mg) of melatonin (MT) vs placebo (PL) on pituitary release of LH, FSH, TSH and PRL after GnRH + TRH and on the adrenocortical release
Effect of a synthetic short-chain ACTH 1-17 (HOE 433) on isoproterenol-induced myocardial infarction in rats.
S Asha Devi
Indian journal of experimental biology, 26(9), 685-687 (1988-09-01)
G P Ceda et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 19(8), 361-363 (1987-08-01)
Recently we demonstrated that ACTH 1-17 infusion in normal subjects is able to stimulate growth hormone (GH) secretion. In order to study the mechanism by which ACTH 1-17 induces this hormonal secretory pattern, we examined the effects of ACTH 1-17

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service