Skip to Content
Merck

Skip To

A1663

Atrial Natriuretic Peptide human

≥97% (HPLC)

Synonym(s):

ANF 1-28, hANF

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C127H203N45O39S3
CAS Number:
Molecular Weight:
3080.44
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(CCCNC(N)=N)NC(=O)C(CC(O)=O)NC(=O)C(CCSC)NC(=O)C(CCCNC(N)=N)NC(=O)CNC(=O)CNC(=O)C(Cc1ccccc1)NC(=O)C(CS)NC(=O)C(CO)NC(=O)C(CO)NC(=O)C(CCCNC(N)=N)NC(=O)C(CCCNC(N)=N)NC(=O)C(CC(C)C)NC(=O)C(N)CO)C(=O)NCC(=O)NC(C)C(=O)NC(CCC(N)=O)C(=O)NC(CO)C(=O)NCC(=O)NC(CC(C)C)C(=O)NCC(=O)NC(CS)C(=O)NC(CC(N)=O)C(=O)NC(CO)C(=O)NC(Cc2ccccc2)C(=O)NC(CCCNC(N)=N)C(=O)NC(Cc3ccc(O)cc3)C(O)=O

InChI

1S/C127H205N45O39S3/c1-9-64(6)99(121(209)150-52-94(182)151-65(7)100(188)155-76(34-35-91(129)179)109(197)167-85(56-174)104(192)149-53-96(184)153-78(43-62(2)3)102(190)148-54-97(185)154-89(60-212)119(207)164-82(48-92(130)180)114(202)169-86(57-175)116(204)163-81(46-67-23-14-11-15-24-67)113(201)158-73(27-18-39-143-125(135)136)107(195)166-84(122(210)211)47-68-30-32-69(178)33-31-68)172-111(199)75(29-20-41-145-127(139)140)159-115(203)83(49-98(186)187)165-110(198)77(36-42-214-8)160-105(193)71(25-16-37-141-123(131)132)152-95(183)51-146-93(181)50-147-103(191)80(45-66-21-12-10-13-22-66)162-120(208)90(61-213)171-118(206)88(59-177)170-117(205)87(58-176)168-108(196)74(28-19-40-144-126(137)138)156-106(194)72(26-17-38-142-124(133)134)157-112(200)79(44-63(4)5)161-101(189)70(128)55-173/h10-15,21-24,30-33,62-65,70-90,99,173-178,212-213H,9,16-20,25-29,34-61,128H2,1-8H3,(H2,129,179)(H2,130,180)(H,146,181)(H,147,191)(H,148,190)(H,149,192)(H,150,209)(H,151,182)(H,152,183)(H,153,184)(H,154,185)(H,155,188)(H,156,194)(H,157,200)(H,158,201)(H,159,203)(H,160,193)(H,161,189)(H,162,208)(H,163,204)(H,164,207)(H,165,198)(H,166,195)(H,167,197)(H,168,196)(H,169,202)(H,170,205)(H,171,206)(H,172,199)(H,186,187)(H,210,211)(H4,131,132,141)(H4,133,134,142)(H4,135,136,143)(H4,137,138,144)(H4,139,140,145)

InChI key

KGZJRXKGJRJKCB-UHFFFAOYSA-N

Gene Information

human ... NPPA(4878)

Application

Atrial natriuretic peptide (ANP) has been used to study its cross reactivity in melanin concentrating hormone (MCH) assay. It has also been used to study its effects in nerve fibres and nerve cell bodies.

Biochem/physiol Actions

Atrial natriuretic peptide (ANP) is synthesized in myoendocrine cells of the heart from which it is released into the circulation. It exerts natriuretic, diuretic, and vasodilatory effects through stimulation of guanylate cyclase-linked NPR-A receptors. It plays an important role in blood volume and blood pressure homeostasis.
Exerts natriuretic, diuretic, and vasodilatory effects through stimulation of guanylate cyclase-linked NPR-A receptors.
Atrial natriuretic peptide (ANP) is a cardiac hormone, which maintains electrolyte homeostasis. It prevents adrenocorticotropic hormone (ACTH) release and arginine vasopressin secretion. ANP functions as a neurotransmitter and modulates pituitary and vegetative functions.

Other Notes

Lyophilized from 0.1% TFA in H2O

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
U5754B5900T6674
assay

≥97% (HPLC)

assay

≥97% (HPLC)

assay

≥97% (HPLC)

assay

≥97% (SDS-PAGE)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder

Gene Information

human ... NPPA(4878)

Gene Information

human ... NPPA(4878)

Gene Information

human ... NPPB(4879)

Gene Information

human ... TNF(7124)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

UniProt accession no.

P01160

UniProt accession no.

P01160

UniProt accession no.

P16860

UniProt accession no.

P01375


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. does it cross react with. mouse 

    1 answer
    1. Although not specifically tested, this material is expected to react with a mouse receptor, as the sequence of this human peptide is identical to the mouse counterpart.

      Helpful?

Reviews

No rating value

Active Filters