Skip to Content
Merck
All Photos(1)

Key Documents

B5900

Sigma-Aldrich

Brain Natriuretic Peptide-32 human

≥97% (HPLC), powder

Synonym(s):

BNP-32

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C143H244N50O42S4
CAS Number:
Molecular Weight:
3464.04
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97% (HPLC)

form

powder

composition

Peptide content, ≥65%

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@@H]1NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCSC)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCNC(N)=N)NC(=O)CNC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](CSSC[C@H](NC(=O)CNC(=O)[C@H](CC(C)C)NC(=O)CNC(=O)[C@H](CO)NC(=O)[C@H](CO)NC(=O)[C@H](CO)NC(=O)[C@H](CO)NC1=O)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](Cc3c[nH]cn3)C(O)=O)NC(=O)CNC(=O)[C@H](CO)NC(=O)CNC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H](NC(=O)[C@H](CCSC)NC(=O)[C@H](CCCCN)NC(=O)[C@@H]4CCCN4C(=O)[C@@H](N)CO)C(C)C

InChI

1S/C143H244N50O42S4/c1-13-76(10)112-137(232)189-99(68-199)131(226)188-98(67-198)130(225)187-97(66-197)129(224)186-96(65-196)117(212)166-59-105(202)169-90(52-72(2)3)114(209)163-61-107(204)171-100(132(227)177-83(32-19-22-44-146)124(219)190-111(75(8)9)136(231)184-91(53-73(4)5)127(222)175-84(34-24-46-159-141(151)152)121(216)174-85(35-25-47-160-142(153)154)122(217)185-94(139(234)235)55-78-57-157-71-167-78)69-238-239-70-101(172-108(205)62-165-116(211)95(64-195)170-106(203)60-162-113(208)87(38-39-103(148)200)181-135(230)110(74(6)7)191-126(221)89(41-51-237-12)179-120(215)82(31-18-21-43-145)180-134(229)102-37-27-49-193(102)138(233)79(147)63-194)133(228)182-92(54-77-28-15-14-16-29-77)115(210)164-58-104(201)168-80(33-23-45-158-140(149)150)118(213)173-81(30-17-20-42-144)119(214)178-88(40-50-236-11)123(218)183-93(56-109(206)207)128(223)176-86(125(220)192-112)36-26-48-161-143(155)156/h14-16,28-29,57,71-76,79-102,110-112,194-199H,13,17-27,30-56,58-70,144-147H2,1-12H3,(H2,148,200)(H,157,167)(H,162,208)(H,163,209)(H,164,210)(H,165,211)(H,166,212)(H,168,201)(H,169,202)(H,170,203)(H,171,204)(H,172,205)(H,173,213)(H,174,216)(H,175,222)(H,176,223)(H,177,227)(H,178,214)(H,179,215)(H,180,229)(H,181,230)(H,182,228)(H,183,218)(H,184,231)(H,185,217)(H,186,224)(H,187,225)(H,188,226)(H,189,232)(H,190,219)(H,191,221)(H,192,220)(H,206,207)(H,234,235)(H4,149,150,158)(H4,151,152,159)(H4,153,154,160)(H4,155,156,161)/t76-,79-,80-,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,110-,111-,112-/m0/s1

InChI key

HPNRHPKXQZSDFX-OAQDCNSJSA-N

Gene Information

human ... NPPB(4879)

Amino Acid Sequence

Ser-Pro-Lys-Met-Val-Gln-Gly-Ser-Gly-Cys-Phe-Gly-Arg-Lys-Met-Asp-Arg-Ile-Ser-Ser-Ser-Ser-Gly-Leu-Gly-Cys-Lys-Val-Leu-Arg-Arg-His

General description

The gene encoding brain natriuretic peptide-32 (BNP-32) is localized on human chromosome 1p36.22. It is implicated in several biological functions such as diuresis, natriuresis, hypotensive action and inhibition of aldosterone secretion. BNP-32 acts as a potential biomarker of high left ventricular-diastolic pressure in patients with symptomatic left ventricular (LV) dysfunction. Elevated expression of this protein is observed in acute myocardial infarction (AMI) patients.

Application

Brain natriuretic peptide-32 (BNP-32) has been used to culture skeletal muscle cells.

Biochem/physiol Actions

Brain natriuretic peptide (type B natriuretic peptide) was originally isolated from brain, but is mainly produced in myoendocrine cells of the heart ventricles from which it is released into the circulation. It is involved in blood pressure control and cardiovascular homeostasis.
Involved in blood pressure control and cardiovascular homeostasis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Edin Begic et al.
PloS one, 14(2), e0212261-e0212261 (2019-02-15)
Alzheimer's disease represents the most common age-related neurodegenerative disorder and a leading cause of progressive cognitive impairment. Predicting cognitive decline is challenging but would be invaluable in an increasingly aging population which also experiences a rising cardiovascular risk. In order
Qingqing Wei et al.
Analytical biochemistry, 573, 1-7 (2019-03-09)
Given that the biological functions of proteins may decrease or even be lost due to degradation by proteases, it is of great significance to identify potential proteases that degrade protein drugs during systemic circulation. In this work, we describe a
Defective natriuretic peptide receptor signaling in skeletal muscle links obesity to type 2 diabetes
Marine C, et al.
Diabetes, db150305-db150305 (2015)
Gabriele Grimm et al.
Journal of applied physiology (Bethesda, Md. : 1985), 122(5), 1249-1254 (2016-12-23)
B-type natriuretic peptide (BNP) is a hormone released by the heart in response to volume load and exerts natriuretic properties. It is clinically used as a diagnostic and prognostic biomarker and investigated as a pharmacological agent in the therapy of
Natriuretic peptide system gene variants are associated with ventricular dysfunction after coronary artery bypass grafting
Amanda AF, et al.
Anesthesiology, 110(4), 738-747 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service