Skip to Content
Merck
All Photos(2)

Key Documents

A1106

Sigma-Aldrich

Anti-ATM antibody,Mouse monoclonal

clone MAT3-4G10/8, purified from hybridoma cell culture

Synonym(s):

Anti-Ataxia-Telangiectasia Mutated

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.45
Pricing and availability is not currently available.

biological source

mouse

Quality Level

recombinant

expressed in mouse cell line

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

MAT3-4G10/8, monoclonal

form

PBS solution

mol wt

antigen ~300 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

concentration

~2 mg/mL

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 0.1-0.2 μg/mL using HEK-293T total cel extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATM(472)
mouse ... Atm(11920)

General description

Monoclonal Anti-ATM (mouse IgG1 isotype) is derived from the hybridoma MAT3-4G10/8 produced by the fusion of mouse myeloma cells (NSO) and splenocytes from BALB/c mice immunized with a peptide.

Specificity

Monoclonal Anti-ATM antibody reacts specifically with mouse and human ATM.

Immunogen

peptide spanning positions 1967-1988 of mouse ATM containing a cysteine at its NH2 terminus coupled to KLH.

Application

Monoclonal Anti-ATM antibody can be used in immunoblotting , ELISA and immunoprecipitation.

Biochem/physiol Actions

Anti-Ataxia-Telangiectasia Mutated (ATM) is responsible for the activation and stabilization of p53 in response to double-strand break (DSB). ATM phosphorylates p53 directly on Ser15 and concomitantly activates other kinases that phosphorylate the same molecule on additional sites. Furthermore, human homolog of double minute 2 (Hdm2) is phosphorylated by ATM on Ser395 and this phosphorylation inhibits Hdm2-mediated degradation of p53.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ATM-dependent phosphorylation of Mdm2 on serine 395: role in p53 activation by DNA damage
Maya R, et al.
Genes & Development, 15(9), 1067-1067 (2001)
Tatiana N Moiseeva et al.
DNA repair, 43, 9-17 (2016-05-29)
We describe a dynamic phosphorylation on serine-1940 of the catalytic subunit of human Pol ε, POLE1, following DNA damage. We also describe novel interactions between POLE1 and the iron-sulfur cluster assembly complex CIA proteins CIAO1 and MMS19. We show that
Jasmin Roohi et al.
Journal of human genetics, 62(5), 581-584 (2017-01-27)
Ataxia-telangiectasia (A-T) is an autosomal recessive chromosome breakage disorder caused by mutations in the ATM gene. Typically, it presents in early childhood with progressive cerebellar dysfunction along with immunodeficiency and oculocutaneous telangiectasia. An increased risk of malignancy is also associated
Phosphorylation of Hdmx mediates its Hdm2- and ATM-dependent degradation in response to DNA damage
Pereg Y, et al.
Proceedings of the National Academy of Sciences of the USA, 102(14), 5056-5061 (2005)
Domenico Delia et al.
Human molecular genetics, 13(18), 2155-2163 (2004-07-23)
Hypomorphic mutations of the MRE11 gene are the hallmark of the radiosensitive ataxia-telangiectasia-like disorder (ATLD). Here, we describe a new family with two affected siblings, ATLD5 and ATLD6, now aged 37 and 36, respectively. They presented with late onset cerebellar

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service