Skip to Content
Merck
All Photos(1)

Key Documents

93800

Sigma-Aldrich

Tylose® MH 300

viscosity 150-450 mPa.s, 2 % in H2O(20 °C)

Synonym(s):

Methyl 2-hydroxyethyl cellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

form

powder

Quality Level

color

white to faint beige

viscosity

150-450 mPa.s, 2 % in H2O(20 °C)

transition temp

flocculation range 60-90 °C (0.5 wt. %)

solubility

H2O: soluble 20 mg/mL

SMILES string

O3[C@H]([C@@H]([C@H]([C@@H]([C@H]3COC)OC)OC)OC)O[C@@H]4[C@H](O[C@H]([C@@H]([C@H]4OC)OC)OC)COC.O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]2[C@H](O[C@H]([C@@H]([C@H]2O)O)O)CO.OCCO

InChI

1S/C20H38O11.C12H22O11.C2H6O2/c1-21-9-11-13(23-3)15(24-4)18(27-7)20(30-11)31-14-12(10-22-2)29-19(28-8)17(26-6)16(14)25-5;13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17;3-1-2-4/h11-20H,9-10H2,1-8H3;3-20H,1-2H2;3-4H,1-2H2/t11-,12-,13-,14-,

InChI key

JTWZVKMWOCBJEV-RXUSYSPUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tylose® MH 300 and Tylose® MH 1000, methyl-hydroxyethyl cellose ethers (MHEC), are water-soluble non-ionic polymers used in materials to provide water retention, binding, thickening, film forming and colloid properties.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Tylose is a registered trademark of SE Tylose GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roland Adden et al.
Analytical chemistry, 78(4), 1146-1157 (2006-02-16)
Hydroxyethylmethyl celluloses (HEMC, DS(Me) 1.46-1.66, DS(HE) = 0.14-0.17) have been analyzed with respect to their methyl and hydroxyethyl pattern in the glucosyl units and along the polymer chain. Methyl groups were located by GLC/MS after direct hydrolysis, reduction, and acetylation
M G Lee
Journal of clinical and hospital pharmacy, 7(1), 67-70 (1982-03-01)
A method is reported for the analysis of prostaglandin PGE2 in methylhydroxyethylcellulose gel. Stability studies using the method have shown that the degradation of PGE in the gel is a first order process at 4 degrees and 25 degrees C.
K M Picker et al.
Pharmaceutical development and technology, 3(1), 31-41 (1998-04-09)
The aim of this research was to investigate the elastic-plastic deformation behavior of the cellulose ethers hydroxypropylmethylcellulose (HPMC), hydroxyethylmethylcellulose (HEMC), and sodium carboxymethylcellulose (NaCMC) at relative humidities (RH) of 38, 57, and 75% and assess how the release of drugs
S Zuleger et al.
International journal of pharmaceutics, 217(1-2), 139-152 (2001-04-09)
The present study deals with controlled drug delivery from hydrocolloid tablets by polymer particle erosion. The influence of excipients and formulation factors on the dissolution behaviour of the methyl hydroxyethyl cellulose (MHEC)-tablets is investigated. Linear drug release with low susceptibility
Su Jeong Ban et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(2), 130-134 (2011-11-01)
The effect of hydroxyethyl methylcellulose (HEMC), in comparison with hydroxypropyl methylcellulose (HPMC), on the body weight and lipid metabolism in mice fed with high fat diet was investigated. The animals were given normal control diet (NC group), high fat diet

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service