Skip to Content
Merck
All Photos(1)

Documents

92637

Sigma-Aldrich

Atto 565-Biotin

Synonym(s):

Biotin-Atto 565

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H57ClN6O10S
Molecular Weight:
921.50
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.32

Assay

90% (HPLC)

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 565 nm; λem 590 nm in 0.1 M phosphate pH 7.0

λ

in ethanol (with 0.1% trifluoroacetic acid)

storage temp.

−20°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashwanth C Francis et al.
Cell host & microbe, 23(4), 536-548 (2018-04-13)
The HIV-1 core consists of capsid proteins (CA) surrounding viral genomic RNA. After virus-cell fusion, the core enters the cytoplasm and the capsid shell is lost through uncoating. CA loss precedes nuclear import and HIV integration into the host genome
Suman Lata et al.
Journal of the American Chemical Society, 128(7), 2365-2372 (2006-02-16)
Labeling of proteins with fluorescent dyes offers powerful means for monitoring protein interactions in vitro and in live cells. Only a few techniques for noncovalent fluorescence labeling with well-defined localization of the attached dye are currently available. Here, we present
Takao Nakata et al.
The Journal of cell biology, 194(2), 245-255 (2011-07-20)
Polarized transport in neurons is fundamental for the formation of neuronal circuitry. A motor domain-containing truncated KIF5 (a kinesin-1) recognizes axonal microtubules, which are enriched in EB1 binding sites, and selectively accumulates at the tips of axons. However, it remains
Marie-Luise Humpert et al.
Proteomics, 12(12), 1938-1948 (2012-05-25)
PTMs of extracellular domains of membrane proteins can influence antibody binding and give rise to ambivalent results. Best proof of protein expression is the use of complementary methods to provide unequivocal evidence. CXCR7, a member of the atypical chemokine receptor
Łukasz Krzemiński et al.
Journal of the American Chemical Society, 133(38), 15085-15093 (2011-08-26)
A combined fluorescence and electrochemical method is described that is used to simultaneously monitor the type-1 copper oxidation state and the nitrite turnover rate of a nitrite reductase (NiR) from Alcaligenes faecalis S-6. The catalytic activity of NiR is measured

Articles

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service